Skip to Content
Merck

Skip To

P5874

PTACH

≥98% (HPLC), solid

Synonym(s):

Cpd 51, S-[6-(4-Phenyl-2-thiazolylcarbamoyl)hexyl] thioisobutyrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H26N2O2S2
CAS Number:
Molecular Weight:
390.56
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

solubility

DMSO: 26 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)C(=O)SCCCCCCC(=O)Nc1nc(cs1)-c2ccccc2

InChI

1S/C20H26N2O2S2/c1-15(2)19(24)25-13-9-4-3-8-12-18(23)22-20-21-17(14-26-20)16-10-6-5-7-11-16/h5-7,10-11,14-15H,3-4,8-9,12-13H2,1-2H3,(H,21,22,23)

InChI key

MDYDGUOQFUQOGE-UHFFFAOYSA-N

Biochem/physiol Actions

HDAC inhibitor; more potent than the majority of HDAC inhibitors except for SAHA (gold standard).

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters