Skip to Content
Merck
All Photos(1)

Documents

P5860

Sigma-Aldrich

Protease from Bacillus sp.

liquid, ≥8 U/g

Synonym(s):

Esperase® 8.0L

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

liquid

specific activity

≥8 U/g

mol wt

20--30 kDa

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Protease is an enzyme used to break down proteins by hydrolyzing peptide bonds. Product P5860 is secreted from Bacillus sp. It is a serine-type protease characterized by excellent perfomance at elevated temperature and pH. Protease is used to degrade proteins, to study protease inhibitors and to study thermal inactivation kinetics. The enzyme has been used to test UV radiation as an environmental friendly, non-thermal technology for sterilizing enzymes used in the food industry.

Biochem/physiol Actions

Protease catabolizes proteins by hydrolysis of peptide bonds. Proteases are inactivated by serine active-site inhibitors, such as phenylmethylsulfonyl fluoride (PMSF) and diisopropylfluorophosphate. Proteases, secreted from Bacillus sp., typically have molecular weights ranging from 20,000 to 30,000. They are typcially stabilized by Ca2+ and have high isoelectric points.

Legal Information

A product of Novozyme corp.
Esperase is a registered trademark of Novozymes Corp.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elodie Lefebvre et al.
Materials science & engineering. C, Materials for biological applications, 82, 210-216 (2017-10-14)
Chronic wound colonization by bacterial biofilms is common and can cause various complications. An anti-biofilm strategy was developed around the co-entrapment of a commercially available antiseptic, PHMB (polyhexamethylene biguanide 4mgmL
D R Durham et al.
Journal of bacteriology, 169(6), 2762-2768 (1987-06-01)
An alkalophilic Bacillus sp., strain GX6638 (ATCC 53278), was isolated from soil and shown to produce a minimum of three alkaline proteases. The proteases were purified by ion-exchange chromatography and were distinguishable by their isoelectric point, molecular weight, and electrophoretic
Effect of UV light on microbial proteases: From enzyme inactivation to antioxidant mitigation.
Lante A, et al.
Innovative Food Science & Emerging Technologies, 17, 130-134 (2013)
Yipeng Zhang et al.
Ultrasonics sonochemistry, 68, 105201-105201 (2020-06-11)
Protein is one of the most abundant natural polymeric materials, but only a few studies on nanoproteins have been conducted. In this paper, a novel approach based on ultrasound-assisted enzymatic hydrolysis was employed for the preparation of microkeratin and nanokeratin
Vadim Volkov et al.
International journal of biological macromolecules, 85, 476-486 (2016-01-13)
The current study describes the in vitro phosphorylation of a human hair keratin, using protein kinase for the first time. Phosphorylation of keratin was demonstrated by (31)P NMR (Nuclear Magnetic Resonance) and Diffuse Reflectance Infrared Fourier Transform (DRIFT) techniques. Phosphorylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service