Skip to Content
Merck
All Photos(1)

Key Documents

P5704

Sigma-Aldrich

Monoclonal Anti-Protein Kinase C (PKC) antibody produced in mouse

clone MC5, ascites fluid

Synonym(s):

Monoclonal Anti-Protein Kinase C, Anti-PKC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
eCl@ss:
32160410
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

MC5, monoclonal

mol wt

antigen 80 kDa

contains

15 mM sodium azide

species reactivity

rat, bovine, mouse, human

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1:500 using rat brain cytosol preparation

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Protein Kinase C (PKC) is a phospholipid and calcium dependent enzyme that regulates intracellular signal transduction. Neuronal PKC modulates several functions such as nerve cell excitability, neuronal plasticity, and release of neurotransmitters. PKC inhibitors have been implicated in diabetic retinopathy . Monoclonal Anti-Protein Kinase C (clone MC5) recognizes an epitope located within the amino acid sequence 296-317 at the hinge region (close to or at the trypsin cleavage site) of PKC . The antibody is specific for PKC in humans, bovines, rats and mice. Binding of the antibody to in vitro purified PKC blocks trypsin-mediated partial proteolysis. Phorbol-ester induced down-regulation of the kinase is inhibited by the introduction of Fab fragment of the antibody into a rodent glioma cell line .
Protein kinase C (PKC) is a 77-90kD protein that belongs to serine/threonine protein kinases family. It has a crucial role in cellular regulation, signal transduction and tumor promotion and oncogenesis. Monoclonal Anti-Protein Kinase C (PKC) antibody can be used for identification and localization of PKC in normal and malignant tissues. It can also be used in immunoblotting (diluted 1:500) using rat brain extracts. Mouse Anti-Protein Kinase C (PKC) antibody reacts specifically with PKC polypeptide (80kD). The product has also shown cross reactivity for rat, bovine, mouse and human.

Specificity

Reacts with the 80 kDa polypeptide of PKC. Binding of the antibody to purified kinase in vitro, blocks partial proteolysis by trypsin. Introduction of Fab fragments of the antibody into a rodent glioma cell line inhibits phorbol-ester-induced down-regulation of the kinase.

Immunogen

purified bovine brain protein kinase C

Application

Monoclonal Anti-Protein Kinase C (PKC) antibody can be used for the localization of protein kinase C by using different immunochemical assays like- dot blot, immunoprecipitation and ELISA. It can also be used as a primary antibody in immunohistochemistry (diluted 1:200) .
Monoclonal Anti-Protein Kinase C (PKC) antibody is suitable for use in immunohistochemistry (frozen sections), immunoblot (80 kD, using bovine brain PKC, extracts of rat glioma and murine NIH 3T3 cell lines), dot blot, ELISA, microarray and immunoprecipitation (rat glioma and human cell extracts/lysates).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zi-Li Yu et al.
International journal of molecular medicine, 37(2), 468-474 (2016-01-09)
Lysophosphatidic acid (LPA) is an efficient, bioactive phospholipid involved in various biological processes. In this study, LPA-induced connective tissue growth factor (CTGF/CCN2) expression and the underlying mechanisms were investigated using the MC3T3-E1 cell line. The MC3T3-E1 cells were stimulated with
Influence of intrathecal injection with dexmedetomidine on the
behavioral ability and analgesic effects on rats with neuropathic
pain and expression of protein kinase C in the spinal dorsal horn
Li X and Zhang W
Experimental and Therapeutic Medicine, 16(5), 3835-3840 (2018)
Lorenzo Cangiano et al.
PloS one, 2(12), e1327-e1327 (2007-12-20)
Hyperpolarization-activated cyclic nucleotide-sensitive (HCN) channels mediate the I(f) current in heart and I(h) throughout the nervous system. In spiking neurons I(h) participates primarily in different forms of rhythmic activity. Little is known, however, about its role in neurons operating with
M I López Gálvez
Current pharmaceutical biotechnology, 12(3), 386-391 (2010-10-14)
To review current data regarding protein Kinase C beta (PKCß) inhibitors and its efficacy in reducing the burden of diabetic retinopathy (DR) and diabetic macular edema (DME). MEDLINE search (1980-2009) and presentations to major meetings. DR and DME have emerged
Akira Hara et al.
Stem cells and development, 17(4), 619-627 (2008-04-09)
In vivo transplantation of undifferentiated embryonic stem (ES) cells can produce teratomas with uncontrolled cell proliferation. Although ES cells may be attractive candidates for human cell-replacement therapy in the future, the major limitation of its application to the therapy is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service