Skip to Content
Merck
All Photos(4)

Key Documents

P5515

Sigma-Aldrich

Prostaglandin E1

≥98% (HPLC), powder, vasodilator 

Synonym(s):

(11α,13E,15S)-11,15-Dihydroxy-9-oxoprost-13-enoic acid, Alprostadil, PGE1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H34O5
CAS Number:
Molecular Weight:
354.48
Beilstein:
5294062
EC Number:
MDL number:
UNSPSC Code:
12352211
eCl@ss:
42020658
PubChem Substance ID:
NACRES:
NA.77

Product Name

Prostaglandin E1, ≥98% (HPLC), synthetic

biological source

synthetic

Assay

≥98% (HPLC)

form

(powder to crystals)

solubility

ethanol: 5 mg/mL

storage temp.

−20°C

SMILES string

O=C1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)[C@H](O)C1

InChI

1S/C20H34O5/c1-2-3-6-9-15(21)12-13-17-16(18(22)14-19(17)23)10-7-4-5-8-11-20(24)25/h12-13,15-17,19,21,23H,2-11,14H2,1H3,(H,24,25)/b13-12+/t15-,16+,17+,19+/m0/s1

InChI key

GMVPRGQOIOIIMI-DWKJAMRDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Prostaglandin E1 has been used:
  • as a platelet inhibitor to analyse its effect on autophagy
  • in platelet adhesion assay
  • to investigate its effect on cAMP production in thyroid glands of normal and growth-retarded mouse in vitro.

Biochem/physiol Actions

Prostaglandin E1 (PGE1) is a hormone synthesized from dihomo-γ-linolenic acid (DGLA) by the action of enzyme prostaglandin synthase. It is a vasodilator and activates adenylate cyclase. It is majorly present in human semen and is a potent platelet aggregation inhibitor. PGE1 infusions is effective in treating limb ischemia and peripheral vascular disease. It is unstable and is hydrolyzed to PGA1 in the lungs. Nanoparticle encapsulation of PGE1 is essential for its delivery at the vascular injury site. It elicits vasodilator effects on the retinal artery and vein in central retinal artery occlusion (CRAO). PGE1 and its synthetic analog misoprostol interact with E-prostanoid receptor 4 (EP4) and elicits protection in chronic myelogenous leukemia (CML).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal and molecular structure of prostaglandin E1
Spek, AL
Acta Crystallographica Section B, Structural Science, 33(3), 816-824 (1977)
Dissection of autophagy in human platelets
Feng W, et al.
Autophagy, 10(4), 642-651 (2014)
Responses of the thyroid gland to TSH and other thyroid stimulators in the growth-retarded (grt) mouse
Kobayashi K, et al.
Zoological Science, 18(7), 955-962 (2001)
Mélanie Abonnenc et al.
Blood transfusion = Trasfusione del sangue, 13(2), 255-264 (2014-11-05)
The Intercept Blood SystemTM (Cerus) is used to inactivate pathogens in platelet concentrates (PC). The aim of this study was to elucidate the extent to which the Intercept treatment modifies the functional properties of platelets. A two-arm study was conducted
Prostaglandins and thromboxanes.
B Samuelsson et al.
Annual review of biochemistry, 47, 997-1029 (1978-01-01)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service