Skip to Content
Merck
All Photos(1)

Documents

P5396

Sigma-Aldrich

Phosphomycin disodium salt

antibacterial MurA inhibitor

Synonym(s):

(−)-(1R,2S)-(1,2-Epoxypropyl)phosphonic acid, Fosfomycin, Phosphonomycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5Na2O4P
Molecular Weight:
182.02
Beilstein:
4604425
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

Quality Level

form

powder

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].C[C@@H]1O[C@@H]1P([O-])([O-])=O

InChI

1S/C3H7O4P.2Na/c1-2-3(7-2)8(4,5)6;;/h2-3H,1H3,(H2,4,5,6);;/q;2*+1/p-2/t2-,3+;;/m0../s1

InChI key

QZIQJIKUVJMTDG-JSTPYPERSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Phosphomycin, also known as Fosfomycin, is an antibiotic produced by Streptomyces fradiae. It is used to treat uncomplicated urinary tract infections. It is used for susceptibility studies of Klebsiella pneumoniae and to study in vitro susceptibility testing procedures for fosfomycin tromethamine.

Biochem/physiol Actions

Antibiotic that concentrates in kidney and bladder; reduces nephrotoxicity and ototoxicity of platinum-containing anti-tumor agents. Fosfomycin is a phosphoenolpyruvate analog that irreversibly inhibits UDP-GlcNAc enolpyruvyl tranferase (MurA), an enzyme involved in bacterial cell wall biosynthesis. As a result, the production of N-acetylmuramic acid, an essential element of the peptidoglycan cell wall, is inhibited.

Other Notes

1g,5g,50g
Keep container tightly closed in a dry and well-ventilated place.Keep in a dry place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Endimiani et al.
Antimicrobial agents and chemotherapy, 54(1), 526-529 (2009-11-11)
In vitro activity of fosfomycin was evaluated against 68 bla(KPC)-possessing Klebsiella pneumoniae (KpKPC) isolates, including 23 tigecycline- and/or colistin-nonsusceptible strains. By agar dilution, 93% of the overall KpKPC were susceptible (MIC(50/90) of 16/64 microg/ml, respectively). The subgroup of 23 tigecycline-
A L Barry et al.
Antimicrobial agents and chemotherapy, 35(6), 1235-1238 (1991-06-01)
Fosfomycin tromethamine (previously fosfomycin trometamol) is an orally administered fosfomycin which may be used for single-dose therapy of uncomplicated urinary tract infections. Fosfomycin tromethamine, norfloxacin, and trimethoprim-sulfamethoxazole inhibited greater than 90% of 352 bacterial isolates representing 25 different species; trimethoprim
Avigail Stokar-Avihail et al.
Cell host & microbe, 25(5), 746-755 (2019-05-10)
Temperate phages can adopt either a lytic or lysogenic lifestyle within their host bacteria. It was recently shown that Bacillus-subtilis-infecting phages of the SPbeta group utilize a peptide-based communication system called arbitrium to coordinate the lysogeny decision. The occurrence of peptide-based
Nikos Fatsis-Kavalopoulos et al.
PLoS biology, 18(9), e3000856-e3000856 (2020-09-18)
Antibiotic combination therapies are important for the efficient treatment of many types of infections, including those caused by antibiotic-resistant pathogens. Combination treatment strategies are typically used under the assumption that synergies are conserved across species and strains, even though recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service