P4922
Phosphate-Citrate Buffer with Sodium Perborate
capsule
Synonym(s):
PCR Buffer with Sodium Perborate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
capsule
pH
4.5-5.5
solubility
water: soluble
Application
Phosphate-citrate buffer with sodium perborate has been used as a substrate buffer in enzyme linked immunosorbent assay (ELISA).
Reconstitution
Empty the contents of one capsule in 100 mL of deionized water and dissolve. The contents of one capsule yields 0.05 M phosphate-citrate buffer, containing 0.03% sodium perborate as a substitute for H2O2, pH 5.0 at 25 °C.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Induced myeloperoxidase activity and related superoxide inhibition during hormone replacement therapy
British Journal of Obstetrics and Gynaecology, 108(5), 474-481 (2001)
Zoo biology, 37(2), 90-97 (2018-04-18)
The female giant panda's estrus is known to be photoperiod sensitive, triggered by increasing day length. A pair of giant pandas was brought to Singapore in September 2012 and exposed to a constant temperature and photoperiod during the first 2
Characterization of a mouse model of allergy to a major occupational latex glove allergen Hev b 5
American Journal of Respiratory and Critical Care Medicine, 167(10), 1393-1399 (2003)
Vaccine, 37(17), 2369-2376 (2019-03-25)
Since the first identification of the H5N1 Goose/Guangdong lineage in 1996, this highly pathogenic avian influenza virus has spread worldwide, becoming endemic in domestic poultry. Sporadic transmission to humans has raised concerns of a potential pandemic and underscores the need
Journal of immunology (Baltimore, Md. : 1950), 204(4), 1022-1034 (2020-01-08)
Abs are glycoproteins that carry a conserved N-linked carbohydrate attached to the Fc whose presence and fine structure profoundly impacts on their in vivo immunogenicity, pharmacokinetics, and functional attributes. The host cell line used to produce IgG plays a major
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service