Skip to Content
Merck
All Photos(1)

Documents

P4875

Sigma-Aldrich

D-Penicillamine

98-101%

Synonym(s):

3,3-Dimethyl-D-cysteine, 3-Mercapto-D-valine, D-(−)-2-Amino-3-mercapto-3-methylbutanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(SH)CH(NH2)CO2H
CAS Number:
Molecular Weight:
149.21
Beilstein:
1722375
EC Number:
MDL number:
UNSPSC Code:
51202303
PubChem Substance ID:
NACRES:
NA.76

Quality Level

Assay

98-101%

form

powder

mp

210 °C (dec.) (lit.)

solubility

H2O: 100 mg/mL

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

CC(C)(S)[C@@H](N)C(O)=O

InChI

1S/C5H11NO2S/c1-5(2,9)3(6)4(7)8/h3,9H,6H2,1-2H3,(H,7,8)/t3-/m0/s1

InChI key

VVNCNSJFMMFHPL-VKHMYHEASA-N

Gene Information

mouse ... Oprk1(18387)

Looking for similar products? Visit Product Comparison Guide

General description

D-Penicillamine is derived from the hydrolysis of the corresponding beta-lactam antibiotic.

Application

It is used as an antirheumatic and as a chelating agent in Wilson′s disease. It is used as a copper chelator to form mixed disulfides with cysteine or other sulfide media components. It is used to inactivate protein-1 DNA binding and to inhibit the growth of asynchronous cultures of rabbit articular chondrocytes.

Biochem/physiol Actions

Penicillamine is a characteristic degradation product of penicillin type antibiotics. One atom of copper combines with two molecules of penicillamine. Penicillamine reduces excess cystine excretion in cystinuria. This is by disulfide interchange between penicillamine and cystine, which results in formation of a readily excreted penicillamine-cysteine disulfide. Penicillamine interferes with the formation of cross-links between tropocollagen molecules and cleaves them when newly formed. Penicillamine lowers IgM rheumatoid factor and depresses T-cell activity.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Jaffray et al.
Annals of the rheumatic diseases, 43(2), 333-338 (1984-04-01)
The long-acting antirheumatic drug D-penicillamine was found to inhibit the growth of asynchronous cultures of rabbit articular chondrocytes. This inhibitory effect was dose-related between 5 X 10(-4) M and 5 X 10(-3) M and was time-dependent for a given dose.
Hao Ren et al.
Microorganisms, 7(12) (2019-12-15)
Probiotics and phytobiotics have been studied as in-feed antibiotic alternatives for decades, yet there are no studies on their possible symbiotic effects. In the present study, newly hatched chickens were fed with feeds supplemented either with host-specific Lactobacillus strains (L.
M L Handel et al.
Molecular pharmacology, 50(3), 501-505 (1996-09-01)
D-Penicillamine (beta, beta-dimethyl cysteine) is an antirheumatic thiol drug with a poorly understood mechanism of action. On the basis that gold(I) thiolates and D-penicillamine are both capable of forming stable bonds with endogenous thiols, we sought a common target of
R John Aitken et al.
Biology of reproduction, 87(5), 110-110 (2012-08-31)
The prolonged incubation of human spermatozoa in vitro was found to induce a loss of motility associated with the activation of mitochondrial reactive oxygen species generation in the absence of any change in mitochondrial membrane potential. The increase in mitochondrial
Wenge Jiang et al.
Nature communications, 10(1), 2318-2318 (2019-05-28)
Since Pasteur first successfully separated right-handed and left-handed tartrate crystals in 1848, the understanding of how homochirality is achieved from enantiomeric mixtures has long been incomplete. Here, we report on a chirality dominance effect where organized, three-dimensional homochiral suprastructures of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service