Skip to Content
Merck
All Photos(3)

Key Documents

P4638

Sigma-Aldrich

Poly-L-ornithine hydrobromide

suitable for cell culture, Mol wt >100,000

Synonym(s):

L-Ornithine homopolymer hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

Product Name

Poly-L-ornithine hydrobromide, mol wt >100,000

form

powder

Quality Level

mol wt

>100,000

technique(s)

cell culture | mammalian: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

[Br-].NC(CCCN)C(=O)O.[H+]

InChI

1S/C5H12N2O2.BrH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H

InChI key

GWRQMKDBBHFVIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Thermodynamic Characterization of Polypeptide Complex Coacervation.: The research investigates the thermodynamic properties of polypeptide complex coacervation, including the role of Poly-L-ornithine hydrobromide in these interactions. This study provides insights into the self-assembly processes of polypeptides, which are crucial for designing new biomaterials (Priftis et al., 2012).

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Ramsay et al.
International journal of pharmaceutics, 210(1-2), 97-107 (2001-02-13)
The impetus to develop non-viral gene delivery vectors has led to examination of synthetic polycationic polymers as plasmid DNA (pDNA) condensing agents. Previous reports have highlighted superiority (up to x 10-fold) in the in-vitro transfection of pDNA complexes formed by
Omaditya Khanna et al.
Journal of biomedical materials research. Part A, 95(2), 632-640 (2010-08-21)
Alginate microcapsules coated with a permselective poly-L-ornithine (PLO) membrane have been investigated for the encapsulation and transplantation of islets as a treatment for type 1 diabetes. The therapeutic potential of this approach could be improved through local stimulation of microvascular
A R Boccaccini et al.
Journal of the Royal Society, Interface, 7 Suppl 5, S581-S613 (2010-05-28)
Electrophoretic deposition (EPD) is attracting increasing attention as an effective technique for the processing of biomaterials, specifically bioactive coatings and biomedical nanostructures. The well-known advantages of EPD for the production of a wide range of microstructures and nanostructures as well
S K Tam et al.
Acta biomaterialia, 7(4), 1683-1692 (2010-12-15)
There is a need for better understanding of the biocompatibility of alginate-polycation microcapsules based on their physicochemical characteristics. Microcapsules composed of alginate with 44% (IntG) or 71% (HiG) guluronate, gelled with calcium (Ca) or barium (Ba) and coated with poly-L-lysine
E Ramsay et al.
Journal of drug targeting, 10(1), 1-9 (2002-05-09)
Gene transfer vectors formed between the cationic polyamino acid, poly-(L)-omithine (PLO) and plasmid DNA (pDNA) have demonstrated superior transfection efficiency (up to x 10-fold) compared to equivalent polylysine-based systems in-vitro. The mechanism(s) underlying this observation remains to be elucidated. We

Articles

Discover L-ornithine, an important amino acid supplement, explore how L-ornithine is used in CHO and iPSC culture, and discover how poly-L-ornithine is used to enhance cell attachment.

Discover L-ornithine, an important amino acid supplement, explore how L-ornithine is used in CHO and iPSC culture, and discover how poly-L-ornithine is used to enhance cell attachment.

Discover L-ornithine, an important amino acid supplement, explore how L-ornithine is used in CHO and iPSC culture, and discover how poly-L-ornithine is used to enhance cell attachment.

Discover L-ornithine, an important amino acid supplement, explore how L-ornithine is used in CHO and iPSC culture, and discover how poly-L-ornithine is used to enhance cell attachment.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service