Skip to Content
Merck
All Photos(3)

Documents

P4142

Sigma-Aldrich

1-Oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine

≥98% (GC), ≥99% (TLC)

Synonym(s):

1-(cis-9-Octadecenoyl)-2-hexadecanoyl-sn-glycero-3-phosphocholine, L-α-Phosphatidylcholine, β-palmitoyl-γ-oleoyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥98% (GC)
≥99% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)O[C@@H](COC(=O)CCCCCCC\C=C/CCCCCCCC)COP(O)(=O)OCC[N](C)(C)C

InChI

1S/C42H83NO8P/c1-6-8-10-12-14-16-18-20-21-23-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43(3,4)5)51-42(45)35-33-31-29-27-25-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3,(H,46,47)/b21-20-

InChI key

ZLPCRCSDGDOSCT-MRCUWXFGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Karagulian et al.
Physical chemistry chemical physics : PCCP, 10(4), 528-541 (2008-01-10)
The ozonolysis of an approximately one monolayer film of 1-oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine (OPPC) on NaCl was followed in real time using diffuse reflection infrared Fourier transform spectrometry (DRIFTS) at 23 degrees C. Matrix-assisted laser desorption/ionization (MALDI) mass spectrometry and Auger electron spectroscopy
A M Jiménez-Monreal et al.
Biochemistry, 38(24), 7747-7754 (1999-07-01)
The activation of protein kinase C alpha was studied by using a lipid system consisting of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocholine (POPC)/1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoserine (POPS) (molar ratio 4:1) and different proportions of 1-palmitoyl-2-oleoyl-sn-glycerol (POG). The phase behavior of the lipidic system was characterized by using differential
Cell biology. Gentle slam.
R Peters et al.
Science (New York, N.Y.), 282(5397), 2213-2214 (1999-01-16)
Mojtaba Bagheri et al.
Antimicrobial agents and chemotherapy, 53(3), 1132-1141 (2008-12-24)
Early studies of immobilized peptides mainly focused upon the relationship between structural properties and the activity of soluble and surface-tethered sequences. The intention of this study was to analyze the influence of immobilization parameters upon the activity profile of peptides.
Jeffrey F Ellena et al.
Journal of medicinal chemistry, 45(25), 5534-5542 (2002-12-03)
There is considerable interest in the use of perfluorocarbons as oxygen carriers in clinical settings; however, little is known regarding the molecular interactions made by these apolar compounds with biological membranes or their effect on membrane structure. NMR spectroscopy was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service