Skip to Content
Merck
All Photos(2)

Key Documents

P3125

Sigma-Aldrich

Papain from papaya latex

buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein

Synonym(s):

Papainase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
42030311
NACRES:
NA.54

form

buffered aqueous suspension

quality

2× Crystallized

specific activity

≥16 units/mg protein

mol wt

23.4 kDa

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Papain, a cysteine protease, consists of a single polypeptide chain with three disulfide bridges and a sulfhydryl group.

Application

Papain from papaya latex has been used to digest:
  • Muller glial cells
  • cartilage explants
  • GAG and DNA content of the tissue constructs
  • decellularized tissue samples.

Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues. Papain has been used to isolate morphologically intact cortical neurons from postnatal rats. Limited papain digestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify the red cell surface to enhance or destroy the reactivity of many red cell antigens. Papain is useful for platelet serology studies. Papain has also been used in the enzymatic synthesis of amino acids, peptides, and other molecules.
Used to produce Fab fragments of antibodies. Also used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.

Biochem/physiol Actions

A cysteine protease that cleaves peptide bonds of basic amino acids, leucine, or glycine.
pH optimum 6.0-7.0
Also hydrolyzes esters and amides.
Papain has broad specificity, in cleaving peptide bonds of basic amino acids, leucinehydrolyzes esters and amides. Papain digests many protein substrates extensively than pancreatic proteases. Papain is commonly used in cell isolation procedures since it is more efficient and less harmful compared to other proteases. Papain significantly increases the yield of viable smooth muscle cells without affecting the cell′s sensitivity to stimulants. It catalyzes several reactions, like hydrolysis, transferase action, specificity and acyl-enzyme intermediate.
Papain papaya latex has antifungal activity against C. albicans. It is a cysteine protease that cleaves peptide bonds of basic amino acids, leucine, or glycine.
. The pH optimum is 6.0-7.0
. Papain hydrolyzes esters and amides. Papain consists of a single polypeptide chain with three disulfide bridges and a sulfhydryl group necessary for activity of the enzyme.

Unit Definition

One unit will hydrolyze 1.0 μmole of N-α-benzoyl-L-arginine ethyl ester (BAEE) per minute at pH 6.2 at 25 °C.

Physical form

Suspension in 0.05 M sodium acetate, pH 4.5, containing 0.01% thymol

Other Notes

View more information on papain at www.sigma-aldrich.com/enzymeexplorer.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTH decreases in vitro human cartilage regeneration without affecting hypertrophic differentiation
Rutgers M, et al.
Testing, 14(4), e0213483-e0213483 (2019)
Scanningless and continuous 3D bioprinting of human tissues with decellularized extracellular matrix
Yu C, et al.
Biomaterials, 194, 1-13 (2019)
Expression of non-visual opsins Opn3 and Opn5 in the developing inner retinal cells of birds. Light-responses in Muller glial cells
Rios MN, et al.
Frontiers in Cellular Neuroscience, 13, 376-376 (2019)
PAPAIN ENZYME: A DIGESTIVE AID
PATEL T
International Journal of Clinical and Biomedical Research, 52-53 (2016)
The dog as a model for osteoarthritis: the FGF4 retrogene insertion may matter
Tellegen AR, et al.
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society (2019)

Articles

Antibody fragmentation with our pepsin digestion protocol for IgG antibody fragmentation and preparation of F(ab’).

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

Uncover properties and applications of the cysteine protease papain and find inhibitors, substrates, and other papain products.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service