Skip to Content
Merck
All Photos(1)

Key Documents

P2459

Sigma-Aldrich

Phenyl-Sepharose 6 Fast Flow

high substitution, extent of labeling: ~40 μmol per mL (high substitution)

Synonym(s):

Phenyl-Agarose

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

form

suspension

extent of labeling

~40 μmol per mL (high substitution)

matrix

agarose, 6% cross-linked

matrix activation

epichlorohydrin

matrix attachment

ether

matrix spacer

3 atoms

particle size

45-165 μm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

P2459-200ML′s updated product is GE17-0973-05

Application

Phenyl Sepharose is used in protein chromatography, affinity chromatography, hydrophobic interaction media, resins and separation media. Phenyl Sepharose has been used in studies that contributed to improving industrial applications in additives in detergents and feed industries. Phenyl Sepharose has also been used to study microbial communities inhabiting hypersaline environments.

Physical form

Suspension in 20% ethanol
aqueous ethanol suspension

Legal Information

Sepharose is a trademark of Cytiva

related product

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K A Rye et al.
The Journal of biological chemistry, 270(1), 189-196 (1995-01-06)
The effect of cholesteryl ester transfer protein (CETP) on the size, composition, and structure of spherical, reconstituted HDL (rHDL) which contain apolipoprotein (apo) A-I as their sole apolipoprotein has been studied. Spherical rHDL were incubated with CETP and Intralipid for
K A Rye et al.
The Journal of biological chemistry, 271(8), 4243-4250 (1996-02-23)
The effect of sphingomyelin (SPM) on the structure and function of discoidal and spherical reconstituted high density lipoproteins (rHDL) has been studied. Three preparations of discoidal rHDL with 1-palmitoyl-2-oleoyl phosphatidylcholine (POPC)/SPM/unesterified cholesterol (UC)/apolipoprotein (apo)A-I molar ratios of 99.6/0. 0/10.2/1.0, 86.0/13.6/10.8/1.0
Frank J Ruzicka et al.
Biochimica et biophysica acta, 1774(2), 286-296 (2007-01-16)
A gene eam in Clostridium difficile encodes a protein that is homologous to lysine 2,3-aminomutase (LAM) in many other species but does not have the lysyl-binding residues Asp293 and Asp330 in LAM from Clostridium subterminale SB4. The C. difficile protein
Development of a downstream process for the isolation of <I>Staphylococcus aureus</I> arsenate reductase overproduced in <I>Escherichia coli</I>.
Messens, J., et al.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 737, 167-178 (2000)
C Mawadza et al.
Journal of biotechnology, 83(3), 177-187 (2000-10-29)
Cellulases produced by two Bacillus strains, CH43 and HR68, isolated from hot springs in Zimbabwe, were purified to homogeneity from culture supernatants. Both enzymes had molecular mass of 40 kDa and isoelectric point of 5.4. The enzymes also resembled each

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service