Skip to Content
Merck
All Photos(1)

Key Documents

P2371

Sigma-Aldrich

Pamidronate disodium salt hydrate

≥95% (NMR), solid

Synonym(s):

3-Amino-1-hydroxy-1-phosphonopropanephosphonic acid disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H9NO7P2Na2 · xH2O
CAS Number:
Molecular Weight:
279.03 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (NMR)

form

solid

color

white

mp

300 °C

solubility

H2O: 28 mg/mL

originator

Novartis

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].NCCC(O)(P(O)([O-])=O)P(O)([O-])=O

InChI

1S/C3H11NO7P2.2Na.H2O/c4-2-1-3(5,12(6,7)8)13(9,10)11;;;/h5H,1-2,4H2,(H2,6,7,8)(H2,9,10,11);;;1H2/q;2*+1;/p-2

InChI key

TVQNUQCYOOJTMK-UHFFFAOYSA-L

Related Categories

Biochem/physiol Actions

Bone resorption inhibitor; inhibitor of farnesyl diphosphate synthase (IC50 = 200 nM).
Pamidronate disodium has the ability to block Wnt and β-catenin signaling, which modulates the osteogenic differentiation in bone marrow mesenchymal stem cells (BMMSCs). It can also reduce bilirubin-impaired apoptosis and helps to develop dentinogenic dysfunction of stem cells from human deciduous teeth.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coralie Leblicq et al.
Pediatric blood & cancer, 60(5), 741-747 (2012-09-25)
Osteonecrosis (ON) is a severe complication of acute lymphoblastic leukemia (ALL) treatments. Recent studies suggest that bisphosphonates might reduce pain and loss of motor function in patients with ON. We assessed the effects of pamidronate compared to standard care in
Uncomplicated vaginal delivery in two consecutive pregnancies carried to term in a woman with osteogenesis imperfecta type I and bisphosphonate treatment before conception.
Chih-Ping Chen et al.
Taiwanese journal of obstetrics & gynecology, 51(2), 305-307 (2012-07-17)
Alexandre Loupy et al.
The Journal of clinical investigation, 122(9), 3355-3367 (2012-08-14)
Tight regulation of calcium levels is required for many critical biological functions. The Ca2+-sensing receptor (CaSR) expressed by parathyroid cells controls blood calcium concentration by regulating parathyroid hormone (PTH) secretion. However, CaSR is also expressed in other organs, such as
Ratna Chatterjee et al.
British journal of haematology, 159(4), 462-471 (2012-09-13)
This study aimed to evaluate bone remodelling disorders in thalassaemia by using pamidronate (PD) infusion with or without hormone replacement therapy (HRT) as a diagnostic-therapeutic tool. In this prospective study, 24 adult thalassaemia major (TM) and 10 thalassaemia intermedia (TI)
Marcin Kos et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 71(6), 1010-1016 (2013-03-16)
Bacterial colonization of the denuded bone in bisphosphonate-related osteonecrosis of the jaw suggests that bisphosphonates increase bacterial adhesion and biofilm formation. This study evaluated the adhesion of gram-positive and gram-negative bacteria on hydroxyapatite coated with pamidronate, one of the most

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service