Skip to Content
Merck
All Photos(2)

Key Documents

P2263

Sigma-Aldrich

Polyethyleneglycol Bisphenol A Epichlorohydrin Copolymer

mol wt 15,000-20,000 Da

Synonym(s):

Polyethylene glycol compound

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C15H16O2·C3H5ClO·(C2H4O)n·H2O]x
CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

form

flakes

Quality Level

mol wt

15,000-20,000 Da

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polyethyleneglycol bisphenol A epichlorohydrin copolymer is a polyethylene glycol compound that has been shown to protect the intestinal epithelium against microbial invasion during stress.

Application

Polyethyleneglycol bisphenol A epichlorohydrin copolymer has been used on glioma cells to compare therapeutic strategies for malignant gliomas.

Preparation Note

Consists of 2 molecules of polyethylene glycol (mol. wt. 7,000-9,000) joined internally through a homobifunctional aromatic spacer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William N Batts et al.
Virus research, 230, 38-49 (2017-01-16)
A novel virus, rainbow trout orthomyxovirus (RbtOV), was isolated in 1997 and again in 2000 from commercially-reared rainbow trout (Oncorhynchus mykiss) in Idaho, USA. The virus grew optimally in the CHSE-214 cell line at 15°C producing a diffuse cytopathic effect;
Shigeru Yamaguchi et al.
Ultrasonics sonochemistry, 18(5), 1197-1204 (2011-01-25)
Sonodynamic therapy is expected to be a novel therapeutic strategy for malignant gliomas. The titanium dioxide (TiO(2)) nanoparticle, a photosensitizer, can be activated by ultrasound. In this study, by using water-dispersed TiO(2) nanoparticles, an in vitro comparison was made between
Shigeru Yamaguchi et al.
Photochemistry and photobiology, 86(4), 964-971 (2010-05-25)
Titanium dioxide (TiO(2)) is thought to be a photocatalytic agent excited by UV light. Our aim was to investigate the photocatalytic antitumor effect of water-dispersed TiO(2) nanoparticles on C6 rat glioma cells and to evaluate the treatment responses by the
Licheng Wu et al.
Gastroenterology, 126(2), 488-498 (2004-02-06)
During stress, erosion of protective intestinal mucus occurs in association with adherence to and disruption of the intestinal epithelial barrier by invading opportunistic microbial pathogens. The aims of this study were to test the ability of a high-molecular-weight polyethylene glycol
Sangheon Han et al.
Biomedical optics express, 10(7), 3472-3483 (2019-07-31)
Gold nanoparticles (AuNPs) below 10 nm in size can undergo renal clearance, which could facilitate their clinical translation. However, due to non-linear, direct relationship between their absorption and size, use of such "ultra-small" AuNPs as contrast agents for photoacoustic imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service