Skip to Content
Merck

Skip To

P1629

TWEEN® 60

nonionic detergent

Synonym(s):

Polyethylene glycol sorbitan monostearate, Polyoxyethylene sorbitan monostearate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

mol wt

estimated mol wt 1309

composition

stearic acid, 40-60% , total stearic and palmitic acid, ≥90%

technique(s)

DNA extraction: suitable, RNA extraction: suitable, cell culture | mammalian: suitable, protein purification: suitable

CMC

27 mg/L

density

1.044 g/mL at 25 °C (lit.)

HLB

14.9

SMILES string

O1C(C(C(C1)OCCO)OCCO)C(OCCO)COCCOC(=O)CCCCCCCCCCC

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

General description

TWEEN 60 is a niosome.

Application

Polyethylene glycol sorbitan monostearate, or TWEEN 60, has been used in a study to assess encapsulation of functional lipophilic components in surfactant-based colloidal delivery systems. [1] It has also been used in a study to investigate doxorubicin loaded magneto-niosomes for targeted drug delivery. [2]

Legal Information

TWEEN is a registered trademark of Croda International PLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
STS02028.170761547947
technique(s)

DNA extraction: suitable, cell culture | mammalian: suitable, RNA extraction: suitable, protein purification: suitable

technique(s)

-

technique(s)

API processing | nano-milling: suitable

technique(s)

-

Quality Level

200

Quality Level

200

Quality Level

500

Quality Level

-

HLB

14.9

HLB

14.9

HLB

-

HLB

-

density

1.044 g/mL at 25 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

density

1.1 g/cm3 at 20 °C

density

-

mol wt

estimated mol wt 1309

mol wt

estimated mol wt 1309

mol wt

-

mol wt

-

CMC

27 mg/L

CMC

27 mg/L

CMC

-

CMC

-


Still not finding the right product?

Explore all of our products under TWEEN® 60


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters