Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.25
UNSPSC Code:
12161700
Pricing and availability is not currently available.
General description
Sigma′s Prehybridization Solution is a 2x ready-to-use buffer for Northern and Southern blot protocols.
Application
Suitable for use as a pre-hybridization buffer in Northern and Southern blot protocols.
Preparation Note
Sigma′s 2X Prehybridization Solution contains 200 μg/mL DNA in 10x SSC and 2X Denhardt′s solutions:
- 200 μg/mL DNA
- 2mg Bovine Serum Albumin (BSA)
- 2mg Ficoll
- 2mg Polyvinylpyrrolidone (PVP)
- 0.15 M sodium citrate
- 1.5M NaCl
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 100, 200 | Quality Level 100 | Quality Level 200 | Quality Level 200 |
| form liquid (2x concentrate) | form liquid (2x concentrate) | form solution | form liquid |
| storage temp. -10 to -25°C | storage temp. −20°C | storage temp. room temp | storage temp. 2-8°C |
| foreign activity DNAses, RNases, NICKases, endonucleases, and exonucleases, none detected | foreign activity DNAses, RNases, NICKases, endonucleases, and exonucleases, none detected | foreign activity - | foreign activity - |
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P1415-5X50ML | 04061834354970 |
| P1415-50ML | 04061834354949 |



