Skip to Content

P0474

L-α-Phosphatidyl-L-serine from Glycine max (soybean)

≥97%

Synonym(s):

1,2-Diacyl-sn-glycero-3-phospho-L-serine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.25
UNSPSC Code:
12352211
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

soybean

Quality Segment

assay

≥97%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

Biochem/physiol Actions

A slowly metabolized structural phospholipid.

Disclaimer

Solutions exposed to room temperature will decompose about 0.5% per day.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library