Skip to Content
Merck
All Photos(1)

Documents

P0370

Sigma-Aldrich

Anti-phospho-Myristoylated Alanine-Rich Protein Kinase C Substrate (pSer152/156) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-phospho-MARCKS (pSer152/156)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 80 kDa

species reactivity

rat, human, mouse

technique(s)

western blot: 1:1000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MARCKS(4082)
mouse ... Marcks(17118)
rat ... Marcks(25603)

General description

The proteins of myristoylated alanine-rich C kinase substrate (MARCKS) family are identified as the ubiquitous substrates of Protein Kinase C (PKC). There are two identified members of MARCKS family, a 32 kDa ubiquitously expressed protein and a 20 kDa protein that is expressed in brain, macrophages and reproductive tissues. MARCKS proteins have vital role to play in the development of brain, cellular migration and adhesion, phagocytosis, neurosecretion and postnatal survival. MARCKS is also a key regulator in the mucin granule release and thereby secretion of mucus in airway. The phosphorylation domain of MARCKS protein contains the serine residues (serine 152, 156 and 163) that are activated by PKC. Besides PKC, MARCKS is also a target for calcium-calmodulin (CaM) and is reported as the potential protein that mediates a crosstalk between CaM and PKC. MARCKS participates in regulation of cytoskeleton by binding with actin and cell membrane
Anti-phospho-MARCKS (pSer152/156) specifically recognizes MARCKS (80 kDa) phosphorylated at serine 152/156.

Immunogen

synthetic phosphorylated peptide derived from the region of rat MARCKS, which is phosphorylated on serine 152/156.

Application

A working dilution of 1:1000 is recommended for detection of MARCKS, phosphorylated at Ser152/156, by immunoblotting in rat hippocampal tissue homogenates.

Physical form

Solution in 10 mM HEPES, pH 7.5, containing 150 mM NaCl, 100 μg/ml BSA and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Arbuzova et al.
The Biochemical journal, 362(Pt 1), 1-12 (2002-02-07)
The proteins of the MARCKS (myristoylated alanine-rich C kinase substrate) family were first identified as prominent substrates of protein kinase C (PKC). Since then, these proteins have been implicated in the regulation of brain development and postnatal survival, cellular migration
The MARCKS brothers: a family of protein kinase C substrates.
A Aderem
Cell, 71(5), 713-716 (1992-11-27)
R H Palmer et al.
FEBS letters, 378(3), 281-285 (1996-01-15)
The 80kDa Myristolated Alanine-Rich C-Kinase Substrate (MARCKS) is a major in vivo substrate of protein kinase C (PKC). Here we report that MARCKS is a major substrate for the lipid-activated PKC-related kinase (PRK1) in cell extracts. Furthermore, PRK1 is shown
Duncan F Rogers
Respiratory care, 52(9), 1134-1146 (2007-08-25)
Mucus secretion is the first-line defense against the barrage of irritants that inhalation of approximately 500 L of air an hour brings into the lungs. The inhaled soot, dust, microbes, and gases can all damage the airway epithelium. Consequently, mucus
The MARCKS family of cellular protein kinase C substrates.
P J Blackshear
The Journal of biological chemistry, 268(3), 1501-1504 (1993-01-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service