Skip to Content
Merck

P0085

Anti-Podoplanin antibody, RAT monoclonal

clone NZ-1.2, purified from hybridoma cell culture

Synonym(s):

Anti-AGGRUS, Anti-GP36, glycoprotein, 36-KD, Anti-GP40, Anti-Gp38, Anti-OTS8, Anti-T1A, Anti-T1A-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
NZ-1.2, monoclonal
Application:
FACS, ICC, IP, WB
Citations:
7
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

Quality Segment

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

NZ-1.2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36 kDa

species reactivity

human

concentration

~1.5 mg/mL

technique(s)

flow cytometry: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: 4-8 μg/mL using U87 total cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDPN(10630)

General description

Monoclonal Anti-Podoplanin (rat IgG2a isotype) is derived from the hybridoma NZ-1.2 produced by the fusion of mouse myeloma cells and splenocytes from CD rat immunized with platelet-aggregation-stimulating (PLAG) domain of podoplanin. Podoplanin (Aggrus) is an integral membrane mucoprotein present on the surface of podocytes in kidney glomeruli and the parietal cells of the glomerular Bowman′s capsule. It is a single membrane-spanning protein composed of a heavily glycosylated extracellular domain and a short intracytoplasmic tail. The extracellular domain possesses a platelet aggregation stimulating (PLAG) domain. Podoplanin has been described to be present in normal, inflammatory and neoplastic lymphatic capillary endothelium.

Immunogen

platelet-aggregation-stimulating (PLAG) domain of human podoplanin.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)
Anti-Podoplanin antibody, Mouse monoclonal has been used in:
  • immunoblotting
  • immunoprecipitation
  • flow cytometry
  • immunocytochemistry

Biochem/physiol Actions

Expression of podoplanin is regulated by the lymphatic-specific homeobox gene Prox-1 (prospero-1). It is involved in lymphatic vessel formation and might be associated with tissue development and repair. Podoplanin is detected in human angiosarcomas, lymphangiomas, and in Kaposi′s sarcomas. It is implicated in the onset of tumor invasion, metastasis or malignant progression. Podoplanin plays an important role in the maintenance of podocyte foot processes and glomerular permeability.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

wgk

WGK 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library