Skip to Content
Merck
All Photos(1)

Key Documents

P0077

Sigma-Aldrich

Prostratin

≥98% (HPLC)

Synonym(s):

12-Deoxyphorbol-13-acetate, CCRIS 6292, NSC 623310

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30O6
CAS Number:
Molecular Weight:
390.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

C[C@@H]1C[C@]2(OC(C)=O)[C@H]([C@@H]3C=C(CO)C[C@@]4(O)[C@@H](C=C(C)C4=O)[C@@]13O)C2(C)C

InChI

1S/C22H30O6/c1-11-6-16-20(26,18(11)25)9-14(10-23)7-15-17-19(4,5)21(17,28-13(3)24)8-12(2)22(15,16)27/h6-7,12,15-17,23,26-27H,8-10H2,1-5H3/t12-,15+,16-,17-,20-,21+,22-/m1/s1

InChI key

BOJKFRKNLSCGHY-HXGSDTCMSA-N

Application

Prostratin has been used to reverse the latency of human immunodeficiency virus-1 (HIV-1) and to quantify the transcriptional activation of the long terminal repeat (LTR) in latently HIV-1-infected Jurkat (J-Lat) cell lines.

Biochem/physiol Actions

Prostratin, an unusual non-tumorigenic phorbol ester, is an activator of protein kinase C (PKC) and also an activator of nuclear factor KB (NF-KB) mediated through activation of the IKKs (IKB kinases).
Prostratin, an unusual non-tumorigenic phorbol ester, is an activator of protein kinase C (PKC) and also an activator of nuclear factor KB (NF-KB) mediated through activation of the IKKs (IKB kinases). Prostratin exhibits potent in vitro activity by inducing HIV expression in latently infected cell lines and primary cells, thus antagonizing HIV latency. Activation of PKC and NF-kB has been proposed as the mechanism of action. Prostratin also inhibits HIV entry into target cells by down-regulating CD4 and CXCR4 receptors.

Features and Benefits

This compound is featured on the PKC page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holly E Johnson et al.
Journal of natural products, 71(12), 2041-2044 (2008-11-15)
Homalanthus nutans, used by Samoan healers to treat hepatitis, produces the antiviral compound 12-deoxyphorbol 13-acetate, prostratin (1). Prostratin is being developed as an adjuvant therapy to clear latent viral reservoirs, the major obstacle to eradication of HIV-AIDS within the human
Marjan Hezareh
Drug news & perspectives, 18(8), 496-500 (2006-01-05)
The persistence of latent reservoirs of human immunodeficiency virus type 1 (HIV-1) represents a major barrier to virus eradication in patients on combination antiretroviral therapy. It has been suggested that treating infected individuals simultaneously with highly active antiretroviral therapy (HAART)
Roberto A Barrero et al.
BMC genomics, 12, 600-600 (2011-12-14)
Euphorbia fischeriana is an important medicinal plant found in Northeast China. The plant roots contain many medicinal compounds including 12-deoxyphorbol-13-acetate, commonly known as prostratin that is a phorbol ester from the tigliane diterpene series. Prostratin is a protein kinase C
Brian A DeChristopher et al.
Nature chemistry, 4(9), 705-710 (2012-08-24)
Bryostatin is a unique lead in the development of potentially transformative therapies for cancer, Alzheimer's disease and the eradication of HIV/AIDS. However, the clinical use of bryostatin has been hampered by its limited supply, difficulties in accessing clinically relevant derivatives
L Yu et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(3-4), 239-244 (2011-10-11)
Chinese jujube (also known as Chinese date) is the fruit of Ziziphus jujuba Mill. (Rhamnaceae). As a famous folk medicine, it is used as antidote in traditional Chinese formula, Shi Zao Decoction, to relieve the drastic inflammatory irritant nature of

Articles

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

Protein kinase C (PKC) is an AGC kinase that phosphorylates serine and threonine residues in many target proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service