OH30428209
Ohaus® Compass Energy Efficient Portable Balance
Model CX621, weighing capacity 620 g, resolution: 0.1 g, External calibration
Synonym(s):
laboratory balance, movable balance, transportable balance, travel balance, weighing balance
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
metal weighing plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30428209
weighing capacity measuring range
620 g, resolution: 0.1 g
weighing pan W × L
14.2 cm × 12.8 cm
battery power
alkaline, 1000 hr life
Looking for similar products? Visit Product Comparison Guide
General description
Designed with energy efficiency in mind, the Compass CX series offers a battery life of up to 1,000 continuous hours, allowing for uninterrupted operation. Featuring a slim, stackable and light-weight design, the CX fits ideally in lab, industrial, education, and even household settings. The streamlined two-button operation means that the balance can be operated easily, and large backlit LCD display enables easy viewing of weighing results.
Application
Weighing, Display Hold
Features and Benefits
Stability indicator, mechanical and software overload/underload protection, low battery indicator, auto shut-off
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service