Skip to Content
Merck
All Photos(3)

Documents

O6881

Sigma-Aldrich

Oxypurinol

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O2
CAS Number:
Molecular Weight:
152.11
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

1 M NaOH: 49.00-51.00 mg/mL, clear to slightly hazy, colorless to yellow (heated NaOH)

SMILES string

O=C1NC(=O)c2cn[nH]c2N1

InChI

1S/C5H4N4O2/c10-4-2-1-6-9-3(2)7-5(11)8-4/h1H,(H3,6,7,8,9,10,11)

InChI key

HXNFUBHNUDHIGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Oxypurinol has been used as a xanthine oxidase inhibitor, to evaluate its effects on hepatocellular carcinoma cell proliferation. It has also been used as a xanthine inhibitor to elucidate its contribution in the induction of hpv16 late gene expression.
Oxypurinol, an allopurinol metabolite, is used as an inhibitor to study the specificity and kinetics of of xanthine oxidase(s). Oxypurinol is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.

Biochem/physiol Actions

Oxypurinol has the ability to stimulate osteoblast differentiation and enhance the formation of bone. It is used to treat gout and hyperuricemia. Oxypurinol is considered as a strong antioxidant. It can prevent the actions of xanthine oxidase (XO) in osteoblasts.
An allopurinol metabolite.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

L K Stamp et al.
Clinical pharmacology and therapeutics, 90(3), 392-398 (2011-07-29)
The treatment of gout requires a lowering of serum urate (SU) levels, and allopurinol is the drug that is most commonly used for this purpose. The objectives of this study were to define the relationships between allopurinol dose on the
Jan B Derks et al.
Pediatric research, 68(5), 374-380 (2010-07-09)
In complicated labor, neonatal outcome may depend not only on the extent of fetal asphyxia and acidosis but also on the effects on the fetal cardiovascular system of reactive oxygen species (ROS) generated during the ischemia-reperfusion (I/R) associated with repeated
Paolo Puddu et al.
Journal of cardiology, 59(3), 235-242 (2012-03-09)
Uric acid is the end product of purine metabolism. Its immediate precursor, xanthine, is converted to uric acid by an enzymatic reaction involving xanthine oxidoreductase. Uric acid has been formerly considered a major antioxidant in human plasma with possible beneficial
Jochen Springer et al.
International journal of cancer, 131(9), 2187-2196 (2012-02-18)
Cachexia is a common co-morbidity in cancer occurring in up to 80% of patients depending on the type of cancer. Uric acid (UA), the end-product of the purine metabolism, is elevated in cachexia due to tissue wasting and upregulated xanthine
Adenosine causes read-through into the late region of the HPV16 genome in a guanosine-dependent manner
Yu H, et al.
Virology, 521(9), 1-19 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service