Skip to Content
Merck
All Photos(1)

Key Documents

O1014

Sigma-Aldrich

Octreotide

≥98% (HPLC)

Synonym(s):

SMS 201-995

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H66N10O10S2
CAS Number:
Molecular Weight:
1019.24
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

C[C@@H](O)[C@@H](CO)NC(=O)[C@@H]1CSSC[C@H](NC(=O)[C@H](N)Cc2ccccc2)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@H](Cc4c[nH]c5ccccc45)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H]([C@@H](C)O)C(=O)N1

InChI

1S/C49H66N10O10S2/c1-28(61)39(25-60)56-48(68)41-27-71-70-26-40(57-43(63)34(51)21-30-13-5-3-6-14-30)47(67)54-37(22-31-15-7-4-8-16-31)45(65)55-38(23-32-24-52-35-18-10-9-17-33(32)35)46(66)53-36(19-11-12-20-50)44(64)59-42(29(2)62)49(69)58-41/h3-10,13-18,24,28-29,34,36-42,52,60-62H,11-12,19-23,25-27,50-51H2,1-2H3,(H,53,66)(H,54,67)(H,55,65)(H,56,68)(H,57,63)(H,58,69)(H,59,64)/t28-,29-,34-,36+,37+,38-,39-,40+,41+,42+/m1/s1

InChI key

DEQANNDTNATYII-OULOTJBUSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Phe-Cys-Phe-Trp-Lys-Thr-Cys-Thr-ol [Disulfide bridge: 2-7]

General description

Octreotide is a somatostatin analogue with D-Phe-Cys-Phe-D-Trp-Lys-ThrCys-Thr-OH amino acid sequence.

Biochem/physiol Actions

Octreotide is three times more potent than the native hormone in inhibiting the secretion of growth hormone glucagon and insulin in vivo. Octreotide regulates serum prolactin levels and resolves galactorrhea or (secondary) amenorrhea in acromegaly patients. Hence, this peptide can be considered as a potent therapeutic for acromegaly treatment.
Somatostatin analog that is 3-times more potent than the native hormone in inhibiting the secretion of growth hormone.

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Calissendorff et al.
Endocrine, 47(1), 169-176 (2013-11-26)
Metastases from ileal neuroendocrine tumors (NETs) to the myocardium are rare and generally seen in patients with widespread metastatic NET disease. The objectives of this investigation were to describe the frequency of intracardiac metastases in ileal NET patients examined by
Octreotide as primary therapy for acromegaly.
Newman CB
The Journal of Clinical Endocrinology and Metabolism, 83(9), 3034-3040 (1998)
Odaly Toffoletto et al.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society, 30-31, 31-36 (2016-09-23)
To extend available dosing options in the treatment of growth hormone deficiency, a comparative pharmacokinetic and pharmacodynamic phase-1 clinical study involving subcutaneous administration of growth hormone was conducted. The test formulation (biosimilar recombinant human growth hormone; r-hGH; Somatotropin) and reference
Animitra Das et al.
The Cochrane database of systematic reviews, (9)(9), CD006388-CD006388 (2010-09-09)
Routine care for chylothorax in neonate includes either conservative or surgical approaches. Octreotide, a somatostatin analogue, has been used for the management of patients with refractory chylothorax not responding to conservative management. To assess the efficacy and safety of octreotide
Anders Sundin
Best practice & research. Clinical gastroenterology, 26(6), 803-818 (2013-04-16)
Neuroendocrine tumours (NETs) comprise a heterogeneous group of neoplasms with very varying clinical expression. A functioning NET, for instance in the pancreas, may be very small and yet give rise to severe endocrine symptoms whereas a patient with a small

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service