Skip to Content

O0141

Obestatin human

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C116H176N32O33
CAS Number:
Molecular Weight:
2546.83
UNSPSC Code:
51111800
NACRES:
NA.32
Biological source:
synthetic
Form:
lyophilized powder
Assay:
≥97% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

sterility

non-sterile

assay

≥97% (HPLC)

form

lyophilized powder

color

white

solubility

deionized water: 1 mg/mL

shipped in

dry ice

storage temp.

−20°C

SMILES string

[nH]1cncc1C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H]4N(CCC4)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)Cc5ccccc5

InChI

1S/C116H176N32O33/c1-13-61(10)95(146-91(158)53-127-113(178)93(59(6)7)147-110(175)81(50-92(159)160)141-108(173)78(46-65-25-18-15-19-26-65)142-112(177)84-28-22-42-148(84)116(181)63(12)130-105(170)80(49-89(123)156)137-98(163)69(118)45-64-23-16-14-17-24-64)115(180)134-70(27-20-21-41-117)101(166)138-76(44-58(4)5)106(171)143-82(54-149)99(164)126-52-90(157)145-94(60(8)9)114(179)135-74(36-40-88(122)155)104(169)139-77(47-66-29-31-68(151)32-30-66)107(172)132-72(34-38-86(120)153)102(167)131-73(35-39-87(121)154)103(168)140-79(48-67-51-125-56-128-67)109(174)144-83(55-150)111(176)133-71(33-37-85(119)152)100(165)129-62(11)97(162)136-75(96(124)161)43-57(2)3/h14-19,23-26,29-32,51,56-63,69-84,93-95,149-151H,13,20-22,27-28,33-50,52-55,117-118H2,1-12H3,(H2,119,152)(H2,120,153)(H2,121,154)(H2,122,155)(H2,123,156)(H2,124,161)(H,125,128)(H,126,164)(H,127,178)(H,129,165)(H,130,170)(H,131,167)(H,132,172)(H,133,176)(H,134,180)(H,135,179)(H,136,162)(H,137,163)(H,138,166)(H,139,169)(H,140,168)(H,141,173)(H,142,177)(H,143,171)(H,144,174)(H,145,157)(H,146,158)(H,147,175)(H,159,160)/t61-,62-,63-,69-,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,93-,94-,95-/m0/s1

InChI key

IXQOGPZNKNSCJR-QQDMDDJXSA-N

Gene Information

human ... GHRL(51738)

General description

Obestatin is a 23-amino acid peptide. It is encoded by the ghrelin gene.

Application

Human obestatin has been used to study its role in differentiation in rat preadipocytes.

Biochem/physiol Actions

Obestatin, an obesity peptide, along with Ghrelin peptides are derived from the same prepropeptide, Ghrelin. In contrast to Ghrelin, Obestatin suppresses food intake, body weight gain, and gastrointestinal motility in rodents.
It does not participate in the release of growth hormone.
Obestatin is an obesity peptide.

Physical form

Lyophilized from 0.1% aqueous trifluoroacetate (TFA).

Other Notes

Lyophilized from 0.1% TFA in H2O


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library