Skip to Content
Merck
All Photos(1)

Key Documents

N7537

Sigma-Aldrich

NSC 663284

≥98% (HPLC), solid

Synonym(s):

6-Chloro-7-(2-morpholin-4-yl-ethylamino)quinoline-5,8-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16ClN3O3
CAS Number:
Molecular Weight:
321.76
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

red

solubility

DMSO: ≥10 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

ClC1=C(NCCN2CCOCC2)C(=O)c3ncccc3C1=O

InChI

1S/C15H16ClN3O3/c16-11-13(18-4-5-19-6-8-22-9-7-19)15(21)12-10(14(11)20)2-1-3-17-12/h1-3,18H,4-9H2

InChI key

BMKPVDQDJQWBPD-UHFFFAOYSA-N

Gene Information

Application

NSC 663284 has been used as a cell division cycle 25 (CDC25) inhibitor to study its effects on the level of kizuna (kiz) dephosphorylation. It has also been used as a control in horseradish peroxidase/phenol red based assay and resazurin-based redox assay.

Biochem/physiol Actions

NSC 663284 is a quinolinediones, which is involved in the inhibition of cell cycle progression at both G1 and G2/M phase. It is also involved in inhibiting the dephosphorylation and activation of cyclin-dependent kinases (Cdks) in vitro and in vivo. NSC 663284 stops the proliferation of various tumor cell lines.
Potent, irreversible, cell permeable and mixed competitive CDC25 phosphatase family inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

470.1 °F

Flash Point(C)

243.4 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kizuna is a novel mitotic substrate for CDC25B phosphatase
Thomas Y, et al.
Cell Cycle, 13(24), 3867-3877 (2014)
Utility of resazurin, horseradish peroxidase, and NMR assays to identify redox-related false-positive behavior in high-throughput screens
Tarnowski M, et al.
Assay and Drug Development Technologies, 16(3), 171-191 (2018)
Inhibition of DUSP13B Activity by NSC 663284
Lee I S, et al.
Bulletin of the Korean Chemical Society,, 33(10), 3505-3507 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service