Skip to Content
Merck

N6760

Nutrient Mixture F-12 Ham

With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

Ham’s F-12

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

technique(s)

cell culture | mammalian: suitable

components

HEPES: no
sodium pyruvate: 0.11 g/L
phenol red: yes
L-glutamine: yes
NaHCO3: no

shipped in

ambient

storage temp.

2-8°C

Application

Developed for growth of primary rat hepatocytes and rat prostate epithelial cells. Also used in a clonal toxicity assay using CHO cells.

Preparation Note

Formulated to contain 10.6 grams of powder per liter of medium.
Supplement with 1.176 g/L sodium bicarbonate.


Still not finding the right product?

Explore all of our products under Nutrient Mixture F-12 Ham


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library