N5377
Methyl nonadecanoate
≥98% (GC)
Synonym(s):
Nonadecanoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)17COOCH3
CAS Number:
Molecular Weight:
312.53
Beilstein:
1788184
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥98% (GC)
form
powder
mp
37-40 °C (lit.)
functional group
ester
lipid type
saturated FAs
shipped in
dry ice
storage temp.
−20°C
SMILES string
CCCCCCCCCCCCCCCCCCC(=O)OC
InChI
1S/C20H40O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-2/h3-19H2,1-2H3
InChI key
BDXAHSJUDUZLDU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Francesca Biandolino et al.
Foods (Basel, Switzerland), 10(2) (2021-03-07)
The effect of cooking (barbecue-grilling, boiling, microwaving, oven cooking and frying) on lipids, fatty acids (FAs) and lipid quality indices of the mussel Mytilus galloprovincialis was investigated. In general, all processing methods significantly (p < 0.05) modified the fatty acid
Niki Alexi et al.
Food chemistry, 301, 125263-125263 (2019-08-05)
The nutritional and sensory quality of a fish fillet is subject to alterations depending on the culinary method used for preparation. The current study aimed to explore the effects of custom culinary preparation methods (steaming, oven-cooking, frying) on the fillet
Jun Cheng et al.
Bioresource technology, 312, 123607-123607 (2020-06-07)
Switchable solvent N, N, N', N'-tetraethyl-1,3-propanediamine (TEPDA) was proposed to extract lipids from wet Nannochloropsis oceanica with a 5% higher extraction efficiency than chloroform-methanol. It was found that TEPDA acted mainly as an organic solvent to soften and dissolve lipids
Thomai Mouskeftara et al.
Metabolites, 11(2) (2021-02-10)
Adiponutrin (patatin-like phospholipase domain-containing 3; PNPLA3), encoded in humans by the PNPLA3 gene, is a protein associated with lipid droplet and endoplasmic reticulum membranes, where it is apparently involved in fatty acid redistribution between triglycerides and phospholipids. A common polymorphism
Stephanie M Fowler et al.
Meat science, 110, 70-75 (2015-07-21)
A hand held Raman spectroscopic device was used to predict intramuscular fat (IMF) levels and the major fatty acid (FA) groups of fresh intact ovine M. longissimus lumborum (LL). IMF levels were determined using the Soxhlet method, while FA analysis
Protocols
Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service