Skip to Content
Merck
All Photos(1)

Key Documents

N5163

Sigma-Aldrich

Anti-NOTCH4 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-INT3, Anti-Notch homolog 4 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~230 kDa
antigen ~80 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:100-1:200
western blot: 1:500-1:1,000

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... NOTCH4(4855)
mouse ... Notch4(18132)
rat ... Notch4(406162)

General description

NOTCH homolog-4 (NOTCH4) is expressed in breast cancer.

Immunogen

synthetic peptide corresponding to residues 1506-1524 of human NOTCH4. This sequence is 89% identical in mouse and rat.

Biochem/physiol Actions

NOTCH homolog-4 (NOTCH4) has an intracellular domain which detaches from the protein on activation. This domain then translocates to the nucleus and modulates transcription of genes. NOTCH4 also activates various pathways and contributes to carcinogenesis. Mutations in the gene encoding the protein may be linked to schizophrenia.

Physical form

solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic variability testing of neurodevelopmental genes in schizophrenic patients.
Terzic T
Journal of Molecular Neuroscience, 56(1), 205-211 (2015)
Cuijuan Qian et al.
Molecular and cellular biochemistry, 401(1-2), 165-174 (2014-12-17)
Gastric cancer (GC) is one of the most common cancers and lethal malignancies in the world. Discovering novel biomarkers that correlate with GC may provide opportunities to reduce the severity of GC. As one of Notch receptor family members in
AKT and 14-3-3 regulate Notch4 nuclear localization.
Ramakrishnan G
Scientific Reports, 5, 8782-8782 (2015)
NOTCH4 is a potential therapeutic target for triple-negative breast cancer.
Nagamatsu I
Anticancer Research, 34(1), 69-80 (2014)
Ana Mafalda Baptista Tadeu et al.
Development (Cambridge, England), 140(18), 3777-3786 (2013-08-09)
The development of the mature epidermis requires a coordinated sequence of signaling events and transcriptional changes to specify surface ectodermal progenitor cells to the keratinocyte lineage. The initial events that specify epidermal keratinocytes from ectodermal progenitor cells are not well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service