Skip to Content
Merck
All Photos(3)

Key Documents

N3908

Sigma-Aldrich

Anti-Nerve Growth Factor Receptor (NGFR p75) antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous solution

Synonym(s):

NGFR Antibody - Anti-Nerve Growth Factor Receptor (NGFR p75) antibody produced in rabbit, Ngfr Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

enhanced validation

functional assay
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry (frozen sections): 1:10,000 using rat brain
indirect immunofluorescence: 1:500 using NGF-treated PC12 cells
microarray: suitable
western blot: 1:5,000 using whole cell extract of NGF-treated PC12 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

rat ... Ngfr(24596)

General description

Nerve growth factor receptor (NGFR) is encoded by the gene mapped to rat chromosome 10. In rats, immunoreactivity of NGFR has been observed in neuronal cell bodies, axons and/or dendrites of cholinergic and non-cholinergic structures of the brain.
Peptide sequence of the antigen is highly conserved in human (single amino acid substitution) and chicken NGFR p75.

Immunogen

synthetic peptide corresponding to the C-terminal of NGFR p75 of rat origin (amino acids 407-425 with N-terminally added lysine) conjugated to KLH.

Application

Anti-Nerve growth factor receptor (NGFR p75) antibody produced in rabbit has been used in:
  • immunoprecipitation
  • immunocytochemistry
  • immunohistochemistry
  • immunoblotting

Biochem/physiol Actions

Neurotrophins (NT) are growth factors that are essential for the development and maintenance of the nervous system. One of the receptors that mediate the effects of NTs is the Nerve Growth Factor Receptor/TNFRSF16 (p75NGFR, p75NTR). NGFR mediates the effects of NGF that are important in the survival and differentiation of neurons and neural crest-derived cells, during embryonic and early postnatal development. Expression of NGFR has been associated with favorable prognosis and survival in breast and pancreatic cancers.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Chen et al.
Medical science monitor : international medical journal of experimental and clinical research, 21, 3608-3615 (2015-11-22)
Alzheimer's disease (AD) is the most common type of dementia. It causes progressive brain disorder involving loss of normal memory and thinking skills. The transplantation of neural stem cells (NSCs) has been reported to improve learning and memory function of
D Rebmann et al.
Journal of orthopaedic surgery and research, 15(1), 270-270 (2020-07-19)
Sensory nerve endings in ligaments play an important role for the proprioceptive function. Clinical trials show that the sense of body position does not fully recover in the knee joint after reconstructive surgery of the ruptured anterior cruciate ligament. The
Myeongjeong Choo et al.
Nature communications, 8(1), 195-195 (2017-08-05)
Elimination of early-formed redundant synapses during postnatal development is essential for functional neural circuit formation. Purkinje cells (PCs) in the neonatal cerebellum are innervated by multiple climbing fibers (CFs). A single CF is strengthened whereas the other CFs are eliminated
Soheila Pourkhodadad et al.
Cell journal, 21(2), 220-228 (2019-03-03)
Traumatic spinal cord injury (SCI) is considered one of the most devastating injuries leading to neuronal disruption. Olfactory ensheathing cells (OECs) and minocycline have been shown to promote locomotor function after spinal cord injury. In this study, we have tested
Berith K Tingaker et al.
Reproductive biology and endocrinology : RB&E, 4, 45-45 (2006-08-30)
The uterus is exposed to changes such as enlargement and distension during pregnancy and labor. In these processes and in the process of cervical ripening, proprioceptive information is likely to be of great importance. Therefore, we wanted to study the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service