Skip to Content
Merck
All Photos(2)

Documents

N3905

Sigma-Aldrich

Nicarbazin

Synonym(s):

N,N′-bis(4-Nitrophenyl)urea compound with 4,6-dimethyl-2-pyrimidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N4O5 · C6H8N2O
CAS Number:
Molecular Weight:
426.38
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

form

powder

antibiotic activity spectrum

parasites

Mode of action

cell membrane | interferes

SMILES string

Cc1cc(C)nc(O)n1.[O-][N+](=O)c2ccc(NC(=O)Nc3ccc(cc3)[N+]([O-])=O)cc2

InChI

1S/C13H10N4O5.C6H8N2O/c18-13(14-9-1-5-11(6-2-9)16(19)20)15-10-3-7-12(8-4-10)17(21)22;1-4-3-5(2)8-6(9)7-4/h1-8H,(H2,14,15,18);3H,1-2H3,(H,7,8,9)

InChI key

UKHWDRMMMYWSFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nicarbazin is a coccidiostat (antiprotozoal agent) in the poultry industry and is a contraceptive for resident Canada geese and waterfowl. It is used to study avian reproduction and is used to interfere with the formation of the vitelline (yolk) membrane in bird eggs. It has been used to study how delivery method can effect plasma levels of 4,4′-dinitrocarbanilide (DNC), which is the active anticoccidial component of Nicarbazin.

Biochem/physiol Actions

Nicarbazin affects avian reproduction by reducing egg production, egg weight, and egg hatchability. The mechanism of action of Nicarbazin is unknown.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G F Mathis et al.
Veterinary parasitology, 121(1-2), 1-9 (2004-04-28)
A 56-day floor pen study was conducted to determine the appropriate time to administer toltrazuril (Baycox) (TOL) for control of coccidiosis in broiler chickens. Litter was seeded with field strains of Eimeria acervulina, Eimeria maxima and Eimeria tenella. On Days
Martin Danaher et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 25(1), 32-40 (2007-10-25)
A survey was carried out on the occurrence of dinitrocarbanilide (DNC), the marker residue for nicarbazin, in poultry produced in Ireland during 2002-2004. Liver (n = 736) and breast muscle samples (n = 342) were tested. DNC residues were found
V Gaudin et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(23), 2358-2362 (2009-04-07)
Nicarbazin, a coccidiostat, is used as a feed additive in poultry but not in laying hens. Feed contamination may however occur resulting in residues being present in eggs. As a Maximum Residue Limit (MRL) does not exist for nicarbazin residues
Virve Hagren et al.
Journal of agricultural and food chemistry, 52(9), 2429-2433 (2004-04-29)
An automated immunoassay for the detection of nicarbazin residues in poultry eggs and liver was developed. The assay was based on a novel all-in-one dry chemistry concept and time-resolved fluorometry. The analyte specific antibody was immobilized into a single microtiter
A-C Huet et al.
Food additives and contaminants, 22(2), 128-134 (2005-04-13)
Nicarbazin and halofuginone have been widely used as coccidiostats for the prevention and treatment of coccidiosis in poultry. It has been shown that accidental cross-contamination of feed can lead to residues of these compounds in eggs and/or muscle. This paper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service