Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NB.15
UNSPSC Code:
24111500
material
clear (blue printed warnings), polypropylene
packaging
pack of 200 ea (bulk bag)
manufacturer/tradename
MTC™ Bio A9002C
W × H
24 in. (61 cm) × 32 in. (81 cm)
Looking for similar products? Visit Product Comparison Guide
General description
These high-temperature autoclavable bags are manufactured from heavy-gauge polypropylene material that withstands autoclaving at temperatures up to 134˚C (270˚F). Their ability to contain is completely unaffected at standard autoclave temperature (121˚C). Each bag has an oversized white marking panel, suitable for use with all types of permanent markers. They are also prominently marked with the words “Hazardous Waste” and the standard “Biohazard” symbol, conforming to the OSHA standard (29CFR1910.1030). Designed to resist stretching and poking, these bags are extremely pliable, leak-resistant and have seamless bottoms for exceptional strength and integrity. Both sizes (12×26 inch and 24×32 inch) are available in clear or opaque “biohazard” red color. They are supplied in units of 200 bags, with 4 inner packs of 50 bags for convenient dispensing and storage.
Autoclave bags, 24x32" (61 x 81.3cm), clear, biohazard, printed, marking area, 200/cs
Features and Benefits
- Heavy gauge polypropylene
- Large white marking areas
- Strong seamless bottom
- Printed biohazard symbol
- Withstands autoclaving at temperatures up to 134˚C (270˚F)
Legal Information
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| MTCA9002C-200EA | 04065271079045 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service