Skip to Content
Merck

MTCA8401B

MTC Bio BenchBin Benchtop Biohazard Bin

capacity 1.5 L

Synonym(s):

bio waste bin, MTC Bio A8401B, laboratory bin, waste canoster, waste receptacle

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

NACRES:
NB.15
UNSPSC Code:
24111500
Manufacturer/tradename:
MTC Bio A8401B
Material:
red (with black Biohazard warning)
Packaging:
pack of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

red (with black Biohazard warning)

packaging

pack of 1 ea

manufacturer/tradename

MTC Bio A8401B

capacity

1.5 L

size

7 in. ( ) × 5 in. × 7 in.

Looking for similar products? Visit Product Comparison Guide

General description

BenchBin is a personal waste container that any researcher can benefit from. Smaller autoclave bags to dispose of biological waste for subsequent processing or forget the autoclave bag and use the BenchBin as a waste container for non-hazardous waste. The red cover is easily removed from the inner basin for easy emptying of contents. Perfect for pipette, tips, microcentrifuge tubes, and gloves, features a push to open/close lid that keeps materials covered and your bench neat.

Features and Benefits

  • Safely dispose of tips, tubes, gloves, etc.
  • Secures and covers hazardous waste
  • Easy push open/close spring lid
  • Compact footprint

Legal Information

BenchBin is a trademark of MTC Bio
MTC is a trademark of MTC Bio

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
MTCA8401B-1EA04065271078963

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service