Skip to Content
Merck

Skip To

MP0030

LookOut® Mycoplasma Elimination Kit

Synonym(s):

Mycoplasma contamination treatment

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

 kit sufficient for 5 mL mycoplasma elimination

Quality Segment

packaging

pkg of 1 kit

storage condition

protect from light

technique(s)

PCR: suitable

application(s)

detection
microbiology

storage temp.

2-8°C

General description

The kit is comprised of a combination of biological agents that reliably and completely eliminate mycoplasma contamination. The initial treatment of this eradication procedure is adequate for mycoplasma elimination in most applications. The second step suppresses and inactivates any remaining mycoplasma using a follow-up antibiotic treatment.

Application

The LookOut® Mycoplasma Elimination Kit has been developed to quickly and efficiently eliminate mycoplasma contamination from cell cultures.
The LookOut® Mycoplasma Elimination Kit is suitable for the elimination of Mollicutes and related organisms (Mycoplasma, Acholeplasma, Spiroplasma, and Entomoplasma) in cell and virus cultures. It is not effective for treating mycoplasma contamination in Chlamydia cultures.

Features and Benefits

  • Minimal cytotoxic properties of the reagents
  • Highly effective
  • Retention of normal cell characteristics

Legal Information

LookOut is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MP0040AL8917MXRPCLKT1
technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

DNA purification: suitable

technique(s)

Rapid Sterility Testing: suitable

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

storage temp.

room temp

usage

 kit sufficient for 5 mL mycoplasma elimination

usage

sufficient for 25 reactions

usage

-

usage

-

application(s)

detection
microbiology

application(s)

detection
microbiology

application(s)

microbiology

application(s)

bioburden testing
environmental
food and beverages
pharmaceutical
sterility testing

packaging

pkg of 1 kit

packaging

pkg of 1 kit

packaging

-

packaging

pkg of 1 kit

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. Is this elimination reagent compatible with serum-free, feeder-free iPSC culture conditions?

    1 answer
    1. This product has not been tested in serum-free, feeder-free induced pluripotent stem cell culture conditions or on stem cells/induced pluripotent stem cells. The effect on pluripotency is currently unknown. No data are available regarding compatibility with these conditions.

      Helpful?

  2. After I dilute my epithelial cells in the media + primary antibiotic, do I not change the media until they reach 80-90% confluency? I usually change the media on the cells every 2-3 days.

    1 answer
    1. The product information sheet protocol for this product instructs to grow the cells in the initial treatment media until 80–90% confluency before passaging. If a media change is necessary for cell health, it is important to include the elimination reagent in the new media.

      Helpful?

  3. Has this been tested in primary cell lines (i.e. skeletal muscle) is there any interference with the normal cell physiology?

    1 answer
    1. The LookOut Mycoplasma Elimination Kit is designed to efficiently eliminate mycoplasma contamination from cell cultures. This kit uses a combination of biological agents to reliably and completely remove mycoplasma contamination. The initial treatment is generally sufficient for mycoplasma elimination in most applications. A second step involves a follow-up antibiotic treatment to suppress and inactivate any remaining mycoplasma. The reagents have minimal cytotoxic properties, and the development of resistant strains is highly unlikely due to the significantly reduced mycoplasma concentration after the initial treatment. Compared to other products for mycoplasma elimination, inactivation, or suppression, the LookOut Mycoplasma Elimination Kit is more effective and has not been shown to cause changes in normal cell characteristics. It is the least cytotoxic method, with overall cytotoxicity demonstrated to be less than 20% across a diverse group of cell lines

      Helpful?

  4. What antibiotics are included in MP0030, the LookOut® Mycoplasma Elimination Kit? Are fluoroquinolones, macrolides, or tetracyclines part of the kit?

    1 answer
    1. The MP0030 product contains two mycoplasma-active antibiotics, with one of them being a fluoroquinolone.

      Helpful?

  5. How does the mechanism of action (MOA) of Product No. MP0030, LookOut® Mycoplasma Elimination Kit, differ from that of BM cyclin?

    1 answer
    1. The key distinction lies in Product No. MP0030, LookOut® Mycoplasma Elimination Kit, which actively eradicates mycoplasma by disrupting the membrane. As a lipopeptide, it identifies cholesterol-rich regions in the membrane (as the concentration of cholesterol in mycoplasma membranes is higher than in eukaryotic cells), creating a pore through which the surrounding medium flows into the mycoplasma, ultimately causing it to burst. In contrast, standard antibiotics simply inhibit metabolic pathways, necessitating the rinsing out of mycoplasma through a medium change. If the process is halted prematurely, any remaining mycoplasma can regrow.

      Helpful?

Reviews

No rating value

Active Filters