Skip to Content
Merck
All Photos(5)

Key Documents

MAK567

Sigma-Aldrich

Pyruvate Dehydrogenase Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

PDH Activity Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116104

usage

sufficient for 100 colorimetric tests

Quality Level

detection method

colorimetric

storage temp.

−20°C

General description

Pyruvate Dehydrogenase (PDH) is a mitochondrial enzyme which has an important role in carbohydrate metabolism. Combining with dihydrolipoyl transacetylase (E2) and dihydrolipoyl dehydrogenase (E3), PDH (E1) forms a well-characterized enzyme complex (PDC). In the presence of NAD+ and CoA, PDH catalyzes the conversion of pyruvate into Acetyl-CoA and CO2. Thus, PDH links the glycolysis metabolic pathway (cytoplasm) to the citric acid cycle (mitochondria). The Pyruvate Dehydrogenase Activity Assay Kit provides a sensitive, simple, fast, direct and convenient way for the detection of PDH activity in various samples, based on a colorimetric method. In the assay, PDH activity changes a colorless developer to a colored product. The amount of the colorimetric (450 nm) product is proportional to the enzymatic activity present in the sample. The kit can detect PDH activity that is lower than 0.1 mU, in a variety of samples.

Application

Pyruvate Dehydrogenase (PDH) Activity Assay Kit has been used in the colorimetric measurement of PDH Activity in biological samples.

Features and Benefits

Sensitive, simple, fast, direct and convenient way for the detection of PDH activity in various biological samples.
Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.

Click here to download a calculator excel file

Suitability

Suitable for measuring PDH Activity in adherent or suspension cells, from an animal or plant source.

Unit Definition

One unit of pyruvate dehydrogenase is the amount of enzyme that will generate 1.0 µmole of NADH per minute at 37 °C at pH=7.5.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Muta. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid intramolecular coupling of active sites in the pyruvate dehydrogenase complex of Escherichia coli: mechanism for rate enhancement in a multimeric structure
Danson M J, et al.
Proceedings of the National Academy of Sciences of the USA, 75(11), 5386-5390 (1978)
Self-assembly and catalytic activity of the pyruvate dehydrogenase multienzyme complex from Bacillus stearothermophilus
Domingo G J, et al.
European Journal of Biochemistry, 266(3), 1136-1146 (1999)
Self-assembly and catalytic activity of the pyruvate dehydrogenase multienzyme complex of Escherichia coli
Bates D L, et al.
Nature, 268(5618), 313-316 (1977)
Structural basis for inactivation of the human pyruvate dehydrogenase complex by phosphorylation: role of disordered phosphorylation loops
Kato M, et al.
Structure (Oxford, United Kingdom), 16(12), 1849-1859 (2008)
A key role for mitochondrial gatekeeper pyruvate dehydrogenase in oncogene-induced senescence
Kaplon J, et al.
Nature, 498(7452), 109-112 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service