Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.21
UNSPSC Code:
41116104
detection method
fluorometric
relevant disease(s)
hematological disorder; cancer
shipped in
wet ice
storage temp.
−20°C
General description
Glucose-6-phosphate (G6P) is a key intermediate for glucose transport into cells. G6P may also be converted to glycogen or starch for storage in the liver and muscles. G6P is utilized by glucose-6-phosphate dehydrogenase (G6PD) to generate the reducing equivalents in the form of NADPH. This is particularly important in red blood cells where G6PD deficiency leads to hemolytic anemia.
The Glucose-6-Phosphate Assay Kit provides a simple, sensitive, and rapid fluorescence-based method for detecting G6P in biological samples such as serum, plasma, urine, as well as in cell culture samples. In the coupled enzyme assay, the G6P concentration is proportionally related to NADPH that is specifically monitored by a fluorogenic NADPH sensor. The detection limit of the G6P Assay Kit is 3 µM G6P in a 100 µL reaction volume.
The Glucose-6-Phosphate Assay Kit provides a simple, sensitive, and rapid fluorescence-based method for detecting G6P in biological samples such as serum, plasma, urine, as well as in cell culture samples. In the coupled enzyme assay, the G6P concentration is proportionally related to NADPH that is specifically monitored by a fluorogenic NADPH sensor. The detection limit of the G6P Assay Kit is 3 µM G6P in a 100 µL reaction volume.
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
10 - Combustible liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
