Skip to Content
Merck

MAK503

Glucose-6-Phosphate Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Glucose-6-Phosphate Quantification Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12352201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

detection method

colorimetric

relevant disease(s)

hematological disorder

shipped in

wet ice

storage temp.

−20°C

General description

Glucose 6-phosphate (G6P) is glucose sugar phosphorylated on carbon 6. Most of the glucose entering cells is phosphorylated to G6P. G6P has three primary fates within the cell. It lies at the start of two major metabolic pathways: glycolysis and the pentose phosphate pathway. In addition to these metabolic pathways, glucose 6-phosphate may also be converted to glycogen or starch for storage.

Application

The kit is suitable for glucose 6-phosphate determination in plasma, serum, tissue, cells, and culture media.

Biochem/physiol Actions

The Glucose 6-Phosphate Assay Kit provides a simple, and automation-ready procedure for measuring G6P concentration. G6P is oxidized by glucose 6-phosphate dehydrogenase and the formed NADPH is coupled to the formazan (WST-8) chromogen. The intensity of the product color, measured at 460 nm, is proportional to the G6P concentration in the sample.

Features and Benefits

  • Linear detection range: 10 to 1000 µM G6P.
  • Compatible with high-throughput handling systems.



Still not finding the right product?

Explore all of our products under Glucose-6-Phosphate Assay Kit


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library