MAK307
Malachite Green Phosphate Assay Kit
sufficient for 2500 colorimetric tests
Synonym(s):
Phosphate Determination Assay Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 2,500 colorimetric tests
detection method
colorimetric
relevant disease(s)
cardiovascular diseases; gastrointestinal diseases
storage temp.
2-8°C
Related Categories
General description
Phosphate is one of the most abundant minerals in the human body and contains about 1 % of total body weight. Phosphate homeostasis is maintained by the several regulators such as parathyroid hormone (PTH), calcitriol and phosphatonins. Maintenance of phosphate homeostasis is essential for various biological processes, such as energy metabolism, cell signaling, regulation of protein synthesis, skeletal development and bone integrity.† An adequate level of phosphate is needed for the process of apoptosis of mature chondrocytes in the growth plate. Phosphate deficiency is associated with the bone pathology and clinical illness.†
Application
Malachite Green Phosphate Assay Kit has been used in phosphate quantification and steady-state kinetics.
Malachite Green Phosphate Assay Kit has been used to measure the liberation of free orthophosphate in the following applications:
- Phosphatase Assays: liberation of phosphate from peptide, protein, or small molecule substrate.
- Lipase Assays: liberation of phosphate from phospholipids
- Nucleoside Triphosphatase Assays: liberation of phosphate from nucleoside triphosphates (ATP, GTP, TTP, CTP etc).
- Quantitation of phosphate in phospholipids, proteins and DNAs, etc.
- Drug Discovery: high-throughput screen for phosphatase inhibitors.
Features and Benefits
Compatible with high-throughput handling systems.
Suitability
Suitable for phosphate determination in a variety of biological and environmental samples.
Principle
Malachite Green is a basic dye belonging to the triphenylmethane group. It is used in biology for the staining of microscopic samples, such as plant tissue, yeast and bacteria. The malachite green method is a sensitive method for the detection of released phosphate.†† The Malachite Green Phosphate Assay Kit is based on quantification of the green complex formed between Malachite Green, molybdate and free orthophosphate. The rapid color formation from the reaction can be conveniently measured on a spectrophotometer (600 - 660 nm) or on a plate reader. The non-radioactive colorimetric assay kits have been optimized to offer superior sensitivity and prolonged shelf life. The assay is simple and fast, involving a single addition step for phosphate determination. Assays can be executed in tubes, cuvettes or multi-well plates. The assays can be conveniently performed in 96 and 384 well plates for high throughput screening of enzyme inhibitors.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
Flash Point(F)
82.4 °F
Flash Point(C)
28 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Edward G.
Synthetic Dyes in Biology, Medicine And Chemistry (2012)
Swetha Ramadesikan et al.
Human molecular genetics, 30(3-4), 198-212 (2021-02-01)
Lowe Syndrome (LS) is a lethal genetic disorder caused by mutations in the OCRL1 gene which encodes the lipid 5' phosphatase Ocrl1. Patients exhibit a characteristic triad of symptoms including eye, brain and kidney abnormalities with renal failure as the
On targeted proteins involved in the neuroprotective effects of lithium citrate.
Torshin I Y, et al.
Neurology, neuropsychiatry, psychosomatics, 9(1), 2017-2017 null
Ilke Sen et al.
Nature communications, 11(1), 138-138 (2020-01-11)
In C. elegans, the conserved transcription factor DAF-16/FOXO is a powerful aging regulator, relaying dire conditions into expression of stress resistance and longevity promoting genes. For some of these functions, including low insulin/IGF signaling (IIS), DAF-16 depends on the protein SMK-1/SMEK
Methods in Plant Cell Biology, 50 (1995)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service