Skip to Content
Merck
All Photos(2)

Key Documents

MAK107

Sigma-Aldrich

LCAT Activity Assay Kit

Supplied by Roar Biomedical, Inc.

Synonym(s):

Lecithin:cholesterol acyltransferase activity assay kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 fluorometric tests

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

cardiovascular diseases

storage temp.

−20°C

Gene Information

human ... LCAT(3931)

General description

The plasma protein lecithin:cholesterol acyltransferase (LCAT) catalyzes the transfer of an acyl group from the sn2 position of phosphatidylcholine to the 3-hydroxyl group of cholesterol resulting in the formation cholesteryl ester. This enzymatic activity occurs on the surface of high density lipoprotein (HDL). The cholesteryl esters formed by LCAT may be packed into the core of HDL.

Application

Applications for this method include high-throughput screening, mechanism of action studies and structure-activity relationship (SAR) work.
LCAT Activity Assay Kit has been used for measuring lecithin-cholesterol acyl transferase activity in plasma samples.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for measuring phospholipase activity of LCAT in plasma or serum.

Principle

The LCAT Activity Assay Kit is a fluorometric assay useful for measuring phospholipase activity of (λEx=390 nm/λEm=470 nm) LCAT. The assay may be validated by inhibition of LCAT with iodoacetate or another spectrally benign (not Ellman′s reagent) LCAT inhibitor.

Kit Components Only

Product No.
Description

  • Substrate Reagent .1 mL

  • READ Reagent 30 mL

  • LCAT Assay Buffer 20 mL

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Akbar Jafarnejad et al.
The Journal of pharmacology and experimental therapeutics, 324(2), 850-857 (2007-11-15)
Diabetes mellitus has been classified as a conformational disease because of changes induced in the structure and function of proteins due to hyperglycemia. In this study, we investigated the effect of high-dose and long-term use of acetyl salicylic acid (ASA)
Anatol Kontush et al.
Arteriosclerosis, thrombosis, and vascular biology, 27(8), 1843-1849 (2007-06-16)
The purpose of this study was to define heterogeneity in the molecular profile of lipids, including sphingomyelin and sphingosine-1-phosphate, among physicochemically-defined HDL subpopulations and potential relevance to antiatherogenic biological activities of dense HDL3. The molecular profile of lipids (cholesteryl esters
Reversal of high fat diet-induced obesity through modulating lipid metabolic enzymes and inflammatory markers expressions in rats.
Uddandrao V V S, et al.
Archives of Physiology and Biochemistry, 1-7 (2018)
Ameliorative potential of gingerol: Promising modulation of inflammatory factors and lipid marker enzymes expressions in HFD induced obesity in rats.
Naidu P B, et al.
Molecular and Cellular Endocrinology, 419, 139-147 (2016)
Daniel J Conklin et al.
Toxicology and applied pharmacology, 243(1), 1-12 (2009-12-26)
Aldehydes such as acrolein are ubiquitous pollutants present in automobile exhaust, cigarette, wood, and coal smoke. Such aldehydes are also constituents of several food substances and are present in drinking water, irrigation canals, and effluents from manufacturing plants. Oral intake

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service