Skip to Content
Merck
All Photos(1)

Documents

M9659

Sigma-Aldrich

4-Methylumbelliferyl N-acetyl-β-D-galactosaminide

≥98% (HPLC), powder

Synonym(s):

4-Methylumbelliferyl 2-acetamido-2-deoxy-β-D-galactopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO8
CAS Number:
Molecular Weight:
379.36
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Methylumbelliferyl N-acetyl-β-D-galactosaminide, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

solubility

pyridine: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

CC(=O)NC1C(O)C(O)C(CO)OC1Oc2ccc3C(C)=CC(=O)Oc3c2

InChI

1S/C18H21NO8/c1-8-5-14(22)26-12-6-10(3-4-11(8)12)25-18-15(19-9(2)21)17(24)16(23)13(7-20)27-18/h3-6,13,15-18,20,23-24H,7H2,1-2H3,(H,19,21)

InChI key

QCTHLCFVVACBSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Methylumbelliferyl N-acetyl-β-ᴅ-galactosaminide has been used as a substrate in the O-GlcNAcase assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reiko Tanaka et al.
Nihon Ishinkin Gakkai zasshi = Japanese journal of medical mycology, 46(2), 125-128 (2005-05-03)
To evaluate a newly developed identification kit, the RID Zyme CAS test for Candida albicans, 1136 C. albicans and 403 non-albicans Candida strains were tested. Distinction of medically important non-albicans strains, with the exception of C. dubliniensis, was obtained. These
Zhiming Shen et al.
Artificial cells, nanomedicine, and biotechnology, 48(1), 656-663 (2020-02-20)
Smoking is an important factor in the pathogenesis of chronic obstructive pulmonary disease (COPD), which is commonly characterised by cellular senescence and inflammation. Recently, miR-200b has emerged as an important target to cure lung disease; however, the function of miR-200b
M F Simpanya
Journal of medical microbiology, 43(3), 230-233 (1995-09-01)
The 4-methylumbelliferyl-N-acetyl-beta-D-galactosaminide (UAG) test was evaluated in parallel with chlamydospore (CHL) and germ-tube (GT) tests for the identification of Candida spp. The UAG test gave 86.6% correct identification of C. albicans and C. tropicalis. Non-C. albicans and non-C. tropicalis yeasts
M T Dalton et al.
Diagnostic microbiology and infectious disease, 12(6), 521-523 (1989-11-01)
Rapid identification of Candida albicans is performed mainly by the germ-tube test. However, recent reports have suggested that up to 5% of C. albicans species can give false negative results. We describe the use of 4-Methylumbelliferyl N-acetyl-beta-D-galactosaminide (4-MAG) conjugate as
H De Boeck et al.
The Journal of biological chemistry, 259(11), 7067-7074 (1984-06-10)
The number of carbohydrate-binding sites of the GalNAc-specific lectin is four per tetramer. The binding parameters of N-acetyl-D-galactosamine and methyl-N-acetyl-alpha-D- galactosaminide , were determined by titrating the perturbation in the absorption spectrum of the protein. For D-galactosides, it was necessary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service