M9418
Cultispher®
-G standard porosity
Synonym(s):
Cell Culture Product
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
Recommended Products
material
gelatin
sterility
non-sterile
bead size
130-380 μm
particle size
130-380 μm
density
1.02-1.04 g/cm3 at 25 °C
Looking for similar products? Visit Product Comparison Guide
General description
Macroporous gelatin-coated microcarrier beads. General purpose beads for anchorage-dependent cells.
Legal Information
Cultispher is a registered trademark of Percell Biolytica AB
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service