Skip to Content
Merck

M8945

Matrix Metalloproteinase-9 human

recombinant, >90% (SDS-PAGE), buffered aqueous solution

Synonym(s):

Gelantinase-B, Gelatinase, 95 kDa, MMP-9

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

>90% (SDS-PAGE)

form

buffered aqueous solution

mol wt

apparent mol wt ~93 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MMP9(4318)

Biochem/physiol Actions

MMP-9 degrades the collagens that make up the extracellular matrix, which has a role in the control of angiogenesis.

Physical form

Solution in Tris-HCl, pH 7.5, Calcium Chloride, Sodium Chloride, and Brij-35

Analysis Note

The biological activity is measured by its ability to cleave a fluorogenic peptide substrate.


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
M8945-10UG04061834063759