Skip to Content
Merck

M8158

Moxalactam sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H18N6Na2O9S
CAS Number:
Molecular Weight:
564.44
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
265-288-4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].COC2(NC(=O)C(C([O-])=O)c1ccc(O)cc1)C3OCC(CSc4nnnn4C)=C(N3C2=O)C([O-])=O

InChI

1S/C20H20N6O9S.2Na/c1-25-19(22-23-24-25)36-8-10-7-35-18-20(34-2,17(33)26(18)13(10)16(31)32)21-14(28)12(15(29)30)9-3-5-11(27)6-4-9;;/h3-6,12,18,27H,7-8H2,1-2H3,(H,21,28)(H,29,30)(H,31,32);;/q;2*+1/p-2

InChI key

GRIXGZQULWMCLU-UHFFFAOYSA-L

General description

Chemical structure: β-lactam


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library