Skip to Content
Merck
All Photos(4)

Key Documents

M7256

Sigma-Aldrich

N,N′-Methylenebisacrylamide

suitable for electrophoresis (after filtration or allowing insolubles to settle)

Synonym(s):

BIS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2=CHCONH)2CH2
CAS Number:
Molecular Weight:
154.17
Beilstein:
1706297
EC Number:
MDL number:
UNSPSC Code:
41105319
PubChem Substance ID:
NACRES:
NB.22

form

powder

technique(s)

electrophoresis: suitable

suitability

suitable for electrophoresis (after filtration or allowing insolubles to settle)

SMILES string

C=CC(=O)NCNC(=O)C=C

InChI

1S/C7H10N2O2/c1-3-6(10)8-5-9-7(11)4-2/h3-4H,1-2,5H2,(H,8,10)(H,9,11)

InChI key

ZIUHHBKFKCYYJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

To be mixed with acrylamide for making polyacrylamide gels for use in protein and nucleic acid electrophoresis.

Other Notes

Cross-linking agent, for the preparation of polyacrylamide (from 01698 Acrylamide puriss.) used as gel in electrophoresis for the separation, analysis and characterization of nucleic acids, proteins and other charged species; A.H. Gordon, Electrophoresis of Proteins in Polyacrylamide and Starch Gels, Vol. 1, Part. I, of "Laboratory Techniques in Biochemistry and Mol. Biology".

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 1B - Muta. 1B - Repr. 2 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Rickwood, B.D. Hames
Gel Electrophoresis of Nucleic Acids: A Practical Approach (1983)
A Bringard et al.
Electrophoresis, 11(10), 802-809 (1990-10-01)
The principle and some applications of multiphasic electrophoresis with diversified spacers are described. The method combines previously published concepts of introducing highly diversified spacers into the stack of an isotachophoretic system with the theory of snow- and telescope-electrophoresis. The link
A J Aho et al.
British journal of anaesthesia, 115(2), 258-266 (2015-07-04)
Concomitantly recorded Bispectral Index® (BIS) and Entropy™ values sometimes show discordant trends during general anaesthesia. Previously, no attempt had been made to discover which EEG characteristics cause discrepancies between BIS and Entropy. We compared BIS and Entropy values, and analysed
Yan Lu et al.
ACS nano, 4(12), 7078-7086 (2010-11-19)
Here, we demonstrate that bimetallic Au-Pt nanorods (NRs) can be grown in situ into thermosensitive core-shell microgel particles by a novel two-step approach. In the first step, Au NRs with an average width of 6.6 ± 0.3 nm and length
Jia Liu et al.
Carbohydrate polymers, 94(1), 539-546 (2013-04-03)
To better use wheat straw and minimize its negative impact on environment, a novel semi-interpenetrating polymer networks (semi-IPNs) superabsorbent resin (SAR) composed of wheat straw cellulose-g-poly (potassium acrylate) (WSC-g-PKA) network and linear polyvinyl alcohol (PVA) was prepared by polymerization in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service