Skip to Content
Merck
All Photos(4)

Documents

M6876

Sigma-Aldrich

D-Mannose 6-phosphate disodium salt hydrate

≥97.0% dry basis (enzymatic)

Synonym(s):

Sodium (2R,3R,4S,5S)-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11Na2O9P · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

wood (birch wood)

Quality Level

Assay

≥97.0% dry basis (enzymatic)

form

powder

impurities

≤1.0% total fructose 6-phosphate and glucose 6-phosphate

color

white to off-white

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

cation traces

Na: 13.2-17.0% (dry basis)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].OC1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H13O9P.2Na.H2O/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5+,6?;;;/m1.../s1

InChI key

UUWJZXLTPORJKW-XTAHGNKUSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Mannose is a monosaccharide.

Application

Mannose has been used in a study to assess the phenotyping of primary human microglia. It has also been used in a study to investigate the identification of Anogeissus latifolia Wallich and analyze refined gum ghatti.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeroen Melief et al.
Glia, 60(10), 1506-1517 (2012-06-29)
Much is still unknown about mechanisms underlying the phenotypical and functional versatility of human microglia. Therefore, we developed a rapid procedure to isolate pure microglia from postmortem human brain tissue and studied their immediate ex vivo phenotype and responses to
Yunpeng Liu et al.
The Journal of organic chemistry, 76(21), 8682-8689 (2011-10-01)
Mannose-6-phosphate (M6P) containing N-linked glycans are the essential targeting signals for hydrolases sorting in eukaryotic cells. To facilitate their structural and binding analyses, a highly efficient and convergent method has been developed to prepare complex N-linked glycans with well-defined M6P
Wei Cheong Ngeow et al.
Journal of anatomy, 219(5), 638-645 (2011-08-05)
Microsurgical repair of transected peripheral nerves is compromised by the formation of scar tissue and the development of a neuroma, thereby limiting the success of regeneration. The aim of this study was to quantify histomorphometrically the structural changes in neural
Petra Tiels et al.
Nature biotechnology, 30(12), 1225-1231 (2012-11-20)
Lysosomal storage diseases are treated with human lysosomal enzymes produced in mammalian cells. Such enzyme therapeutics contain relatively low levels of mannose-6-phosphate, which is required to target them to the lysosomes of patient cells. Here we describe a method for
Synthesis of mannose-6-phosphate analogues and their utility as angiogenesis regulators.
Véronique Barragan-Montero et al.
ChemMedChem, 6(10), 1771-1774 (2011-07-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service