Skip to Content
Merck
All Photos(2)

Key Documents

M5760

Sigma-Aldrich

4-(4-Methoxybenzylamino)-7-nitrobenzofurazan

>97.5% purity (HPLC), powder

Synonym(s):

4-(4-Methoxybenzylamino)-7-nitro-2,1,3-benzoxadiazole, MBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N4O4
CAS Number:
Molecular Weight:
300.27
Beilstein:
1151390
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

4-(4-Methoxybenzylamino)-7-nitrobenzofurazan,

Assay

>97.5% (HPLC)

Quality Level

form

powder

color

orange to red

solubility

DMSO:methanol (1:1): 50 mg/mL, clear, orange

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

COc1ccc(CNc2ccc([N+]([O-])=O)c3nonc23)cc1

InChI

1S/C14H12N4O4/c1-21-10-4-2-9(3-5-10)8-15-11-6-7-12(18(19)20)14-13(11)16-22-17-14/h2-7,15H,8H2,1H3

InChI key

IENONFJSMWUIQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fluorescent probe for hydrophobic (non-polar) regions in proteins and nucleoproteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moayyad E Suleiman et al.
The British journal of radiology, 91(1085), 20180032-20180032 (2018-02-06)
This work proposes the use of mammographic breast density (MBD) to estimate actual glandular dose (AGD), and assesses how AGD compares to mean glandular dose (MGD) estimated using Dance et al method. A retrospective sample of anonymised mammograms (52,405) was
Shingo Fukuma et al.
Scientific reports, 9(1), 11301-11301 (2019-08-07)
Achieving calcium, phosphorus, and intact parathyroid hormone (PTH) targets helps improve dialysis population outcomes. We aimed to assess the population-level associations of achievement of those targets with survival using population-attributable-fractions (PAFs). We conducted a case-cohort study using data from 8229
Angharad R Morgan et al.
Alzheimer's & dementia : the journal of the Alzheimer's Association, 15(6), 776-787 (2019-05-03)
Plasma biomarkers for Alzheimer's disease (AD) diagnosis/stratification are a "Holy Grail" of AD research and intensively sought; however, there are no well-established plasma markers. A hypothesis-led plasma biomarker search was conducted in the context of international multicenter studies. The discovery
Alessia Armezzani et al.
Development (Cambridge, England), 145(11) (2018-05-10)
The shoot apical meristem of higher plants continuously generates new tissues and organs through complex changes in growth rates and directions of its individual cells. Cell growth, which is driven by turgor pressure, largely depends on the cell walls, which
Shin Miura et al.
Surgical endoscopy, 29(7), 1862-1870 (2014-10-04)
Single preoperative biliary drainage for malignant perihilar biliary stricture occasionally fails to control jaundice and cholangitis. Multiple biliary drainage is required in such cases, but their clinical background is unclear. We determined the clinical characteristics associated with the requirement for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service