Merck
M3907
Metabolic Pathways, 22nd Ed.
Poster
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.28
UNSPSC Code:
41100000
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
publication info
D.E. Nicholson, Sigma Chemical Co., St. Louis, MO, 2003
size
26 in. × 39.5 in.
General description
This 22nd edition of the IUBMB-Nicholson Metabolic Pathways Chart features an enhanced ATP metabolism in mitochondria and chloroplasts. It illustrates the sequences of reactions involved in different aspects of metabolism and how they are integrated. The backbone of the map is the Glycolytic Pathway, followed by the TCA (Krebs) Cycle and the ElectronTransport System, which together produce most of the energy (ATP) required for all life. Most other aspects of metabolism are associated with this backbone and are differentiated by the use of color. Human metabolism is distinguished by the use of black arrows. Some 550 reactions are identified by their IUBMB Enzyme Commission (EC) numbers which are then indexed.
Still not finding the right product?
Explore all of our products under Metabolic Pathways, 22nd Ed.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Global Trade Item Number
SKUGTIN
M3907-1EA04061824138788