Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H17ClN2O2
CAS Number:
Molecular Weight:
268.74
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Product Name
Moclobemide, ≥98% (HPLC), solid
assay
≥98% (HPLC)
form
solid
color
white
solubility
DMSO: >20 mg/mL
originator
Roche
storage temp.
room temp
SMILES string
Clc1ccc(cc1)C(=O)NCCN2CCOCC2
InChI
1S/C13H17ClN2O2/c14-12-3-1-11(2-4-12)13(17)15-5-6-16-7-9-18-10-8-16/h1-4H,5-10H2,(H,15,17)
InChI key
YHXISWVBGDMDLQ-UHFFFAOYSA-N
Biochem/physiol Actions
Moclobemide acts as a substrate for cytochrome P450 2C19 (CYP2C19). It also behaves as an inhibitor of cytochrome P450 2D6 (CYP2D6), cytochrome P450 1A2 (CYP1A2 and CYP2C19. Moclobemide is also involved in the increased expression of B-cell lymphoma 2 (BcL-2). It also causes neural stem cell (NSC) differentiation into serotoninergic neuron by extracellular‐regulated kinase (ERK) pathway.
Moclobemide is a reversible monoamine oxidase A inhibitor (MAOI); antidepressant.
Moclobemide is a reversible monoamine oxidase A inhibitor (MAOI); antidepressant. Elimination half-life in humans = 1 -3 hrs; absolute oral bioavailability. Unlike other MAO inhibitors, does not significantly increase blood pressure in humans upon combination with tyramine.
Features and Benefits
This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

