Skip to Content
Merck

M2378

Mucin from porcine stomach

Type II

Synonym(s):

MUC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
282-010-7
NACRES:
NA.61
MDL number:
Form:
powder
Biological source:
Porcine stomach
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine stomach

Quality Segment

type

Type II

form

powder

composition

bound sialic acid, ≤1.2%

technique(s)

cell culture | mammalian: suitable

solubility

1 M NaOH: soluble 20 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

General description

Mucin is majorly composed of carbohydrate units, crucial for structural architecture and conformation. The monomer of mucin is 640 kDa, with protein contributing to only 20% of the molecular weight. The carbohydrate component is attached to the threonine and serine side chains. Mucins are classified into secreted and transmembrane mucins.
Mucins or mucus glycoproteins are the main macromolecular components of mammalian mucus. It was also used in a study to evaluate the sputum smears concentrated by cyto-centrifugation for detection of acid-fast bacilli.

Application

Mucin from porcine stomach has been used:
  • in the standard curve generation for quantifying mucin-2 generated during mucosal inflammation in mice
  • as a component of digestion fluid solutions in in vitro human digestion model
  • as a component of simulated saliva fluid

Mucin from porcine stomach was used in studies on the binding site of the galactose-specific agglutinin PA-IL from Pseudomonas aeruginosa.

Biochem/physiol Actions

Mucin has wide pharmacological applications in drug delivery. Mucins play a protective role in inflammation. Deregulation in mucin synthesis contributes to tumorigenesis and chronic inflammation.
Mucus forms the protective cover for all epithelial surfaces. The major structural component of mucus is gel-forming mucins. The mucus layer covering the intestinal epithelium has the MUC2 mucin as its central molecule. MUC2 is produced by goblet cells.


Still not finding the right product?

Explore all of our products under Mucin from porcine stomach


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library