Skip to Content
Merck
All Photos(1)

Key Documents

M213

Sigma-Aldrich

Mastoparan 17

≥96.5%, lyophilized powder

Synonym(s):

Mas 17

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H132N20O15
CAS Number:
Molecular Weight:
1493.92
MDL number:
UNSPSC Code:
41106300
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥96.5%

form

lyophilized powder

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](N)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CC(C)C)C(N)=O

InChI

1S/C70H132N20O15/c1-15-41(10)56(76)70(105)90-54(36-55(75)91)69(104)89-52(34-39(6)7)67(102)85-47(25-17-21-29-72)63(98)80-44(13)59(94)82-46(24-16-20-28-71)62(97)79-42(11)58(93)78-43(12)61(96)87-51(33-38(4)5)66(101)81-45(14)60(95)83-48(26-18-22-30-73)64(99)84-49(27-19-23-31-74)65(100)88-53(35-40(8)9)68(103)86-50(57(77)92)32-37(2)3/h37-54,56H,15-36,71-74,76H2,1-14H3,(H2,75,91)(H2,77,92)(H,78,93)(H,79,97)(H,80,98)(H,81,101)(H,82,94)(H,83,95)(H,84,99)(H,85,102)(H,86,103)(H,87,96)(H,88,100)(H,89,104)(H,90,105)/t41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,56-/m0/s1

InChI key

TUCDQOGRFHOSKG-ZWFWRLIFSA-N

Amino Acid Sequence

Ile-Asn-Leu-Lys-Ala-Lys-Ala-Ala-Leu-Ala-Lys-Lys-Leu-Leu-NH2

Biochem/physiol Actions

Inactive analog of mastoparan; useful as negative control.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C De Lisle et al.
Pancreas, 10(4), 374-381 (1995-05-01)
Constitutive membrane trafficking events are regulated by heterotrimeric G-proteins (G-proteins) in addition to their regulation by small GTP-binding proteins (smgs). Here, we used streptolysin O-permeabilized mouse pancreatic acini and compounds that interact with G-proteins, but not smgs, to examine whether
T Higashijima et al.
The Journal of biological chemistry, 265(24), 14176-14186 (1990-08-25)
Mastoparan (MP), a cationic, amphiphilic tetradecapeptide, stimulates guanine nucleotide exchange by GTP-binding regulatory proteins (G proteins) in a manner similar to that of G protein-coupled receptors. 1) MP stimulated exchange by isolated G protein alpha subunits and alpha beta gamma
N Vitale et al.
The Biochemical journal, 300 ( Pt 1), 217-227 (1994-05-15)
We have previously described that mastoparan, an amphiphilic tetradecapeptide that activates heterotrimeric G-proteins, inhibits Ca(2+)-induced MgATP-dependent secretion from streptolysin-O-permeabilized chromaffin cells [Vitale, Mukai, Rouot, Thiersé, Aunis and Bader (1993) J. Biol. Chem. 268, 14715-14723]. Our observations suggest the involvement of
Philip Kitchen et al.
Cancer research, 80(4), 757-770 (2019-12-18)
Aberrant Notch and Wnt signaling are known drivers of cholangiocarcinoma (CCA), but the underlying factors that initiate and maintain these pathways are not known. Here, we show that the proline-rich homeodomain protein/hematopoietically expressed homeobox (PRH/HHEX) transcription factor forms a positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service