Skip to Content
Merck
All Photos(1)

Documents

M0821

Sigma-Aldrich

Moexipril hydrochloride

powder, ≥98% (HPLC)

Synonym(s):

(3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H35N2O7Cl
CAS Number:
Molecular Weight:
535.03
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

solubility

H2O: 32 mg/mL

originator

UCB Inc.

storage temp.

2-8°C

SMILES string

Cl.CCOC(=O)[C@H](CCc1ccccc1)N[C@@H](C)C(=O)N2Cc3cc(OC)c(OC)cc3C[C@H]2C(O)=O

InChI

1S/C27H34N2O7.ClH/c1-5-36-27(33)21(12-11-18-9-7-6-8-10-18)28-17(2)25(30)29-16-20-15-24(35-4)23(34-3)14-19(20)13-22(29)26(31)32;/h6-10,14-15,17,21-22,28H,5,11-13,16H2,1-4H3,(H,31,32);1H/t17-,21-,22-;/m0./s1

InChI key

JXRAXHBVZQZSIC-JKVLGAQCSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Moexipril is an ACE-Inhibitor; Antihypertensive; Unlike other ACE inhibitors, prolongs QT interval leading to torsades de pointes and cardiad arrhythmia;

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by UCB Inc.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mila Volchek et al.
Human reproduction (Oxford, England), 25(10), 2455-2464 (2010-08-24)
The mammalian placenta plays a central role in maternal tolerance of the semi-allogeneic fetus and fluid balance between the maternal and fetal compartments. The lymphatics play a role in both these function. The aim of this study was to describe
Thomas S Jacques et al.
The EMBO journal, 29(1), 222-235 (2009-11-21)
It has been suggested that intrinsic brain tumours originate from a neural stem/progenitor cell population in the subventricular zone of the post-natal brain. However, the influence of the initial genetic mutation on the phenotype as well as the contribution of
K E Kristensen et al.
Clinical pharmacology and therapeutics, 96(6), 713-722 (2014-09-16)
Clopidogrel is an oral antiplatelet prodrug, the majority of which is hydrolyzed to an inactive metabolite by hepatic carboxylesterase 1 (CES1). Most angiotensin-converting enzyme inhibitors (ACEIs) are also metabolized by this enzyme. We examined the effects of ACEIs on clopidogrel

Articles

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service