Skip to Content
Merck
All Photos(3)

Key Documents

M0534

Sigma-Aldrich

5-Methyl-DL-tryptophan

tryptophan analog

Synonym(s):

5-Methyltryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
Beilstein:
20225
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥97% (TLC)

form

powder

color

white to faint yellow

mp

280-282 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cc1ccc2[nH]cc(CC(N)C(O)=O)c2c1

InChI

1S/C12H14N2O2/c1-7-2-3-11-9(4-7)8(6-14-11)5-10(13)12(15)16/h2-4,6,10,14H,5,13H2,1H3,(H,15,16)

InChI key

HUNCSWANZMJLPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Amelioration of colitis through modulation of gut microbiota: The metabolite 5-Methyl-ᴅʟ-tryptophan (5-MT), derived from Angelica sinensis polysaccharides, was found to ameliorate colitis by modulating gut microbiota and the TLR4/MyD88/NF-κB signaling pathway. This suggests a potential application of 5-MT in inflammatory bowel disease research and therapy (Zou et al., 2023).

  • Enzymatic synthesis of tryptophan derivatives: A study on the one-pot enantioselective synthesis of (S)-spirobrassinin and non-natural (S)-methylspirobrassinin from amino acids highlights a method using a turnip enzyme. This research outlines a novel approach to synthesizing tryptophan derivatives, potentially useful in biochemical assays (Ryu et al., 2021).

Biochem/physiol Actions

5-Methyl-DL-tryptophan inhibits the synthesis of anthranilate compounds that are the first steps in the biosynthesis of tryptophan in Neurospora crassa. 5-Methyl-DL-tryptophan is a corepressor of the E. coli trp repressor.
5-Methyl-DL-tryptophan may be used to select genetic mutants of PS strain of Methanococcus voltae (archaebacteria). 5-Methyl-tryptophan is a repressor trp operon expression. 5-Methyl-tryptophan is a substrate for tryptophanase. 5-Methyl-tryptophan inhibits the induction of anthranilate synthase activity by elicitors in oats.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E I Hyde et al.
European journal of biochemistry, 201(3), 569-579 (1991-11-01)
The Escherichia coli trp repressor binds to the trp operator in the presence of tryptophan, thereby inhibiting tryptophan biosynthesis. Tryptophan analogues lacking the alpha-amino group act as inducers of trp operon expression. We have used one- and two-dimensional 1H-NMR spectroscopy
Vered Tzin et al.
The New phytologist, 194(2), 430-439 (2012-02-03)
The shikimate pathway of plants mediates the conversion of primary carbon metabolites via chorismate into the three aromatic amino acids and to numerous secondary metabolites derived from them. However, the regulation of the shikimate pathway is still far from being
Tetsuya Matsukawa et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 57(1-2), 121-128 (2002-04-03)
Oat phytoalexins, avenanthramides, are a series of substituted hydroxycinnamic acid amides with anthranilate. The anthranilate in avenanthramides is biosynthesized by anthranilate synthase (AS, EC 4.1.3.27). Induction of anthranilate synthase activity was investigated in oat leaves treated with oligo-N-acetylchitooligosaccharide elicitors. AS
Y Tozawa et al.
Plant physiology, 126(4), 1493-1506 (2001-08-14)
Anthranilate synthase (AS) is a key enzyme in the synthesis of tryptophan (Trp), indole-3-acetic acid, and indole alkaloids. Two genes, OASA1 and OASA2, encoding AS alpha-subunits were isolated from a monocotyledonous plant, rice (Oryza sativa cv Nipponbare), and were characterized.
H J Cho et al.
Plant physiology, 123(3), 1069-1076 (2000-07-13)
A cDNA clone that encodes a feedback-insensitive anthranilate synthase (AS), ASA2, isolated from a 5-methyl-tryptophan (Trp) (5MT)-resistant tobacco cell line under the control of the constitutive cauliflower mosaic virus 35S promoter, was introduced into the forage legume Astragalus sinicus by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service